Petrophysics is about translating wireline logs into properties related to reservoir formation quality. These properties include static properties, as well as flow properties and saturation of fluids. Digital Rock Analysis (DRA) allows petrophysisists to calculate parameters on existing rock samples that will help with formation evaluation.
Petrophysical analysis software
PerGeos Software is a comprehensive Digital Rock Analysis solution that provides petrophysicists and geophysicists with advanced analytical tools to calculate parameters on existing rock samples, to understand the reservoir formations.
Using whole-core CT data: Generate pseudo-logs. Co-visualize Core CT image, wireline logs, unwrapped Core image. Use Dual-Energy CT (DECT) technique to characterize Core density and Atomic Number or to perform Core classification based on reference materials.
From plugs micro-CT or FIB-SEM imaging data: calculate Absolute Permeability. Run Fluid Flow simulations. Calculate Formation Factor, Electrical Resistivity. Simulate Mercury Injection Capillary Pressure.
Perform multi-scale geological and petrophysical analysis of your core imaging data
From Core to Pore, using imagery from large CT to FIB/SEM systems, PerGeos Software helps you digitally understand and validate observations and properties of your rock data with a high level of confidence.
Ability to interpret images at multiple resolutions from multiple modalities, advanced 2D/3D and 3D/3D registration tools and adaptive thresholding make PerGeos Software a unique digital platform for powerful up-scaling workflows.
This allows geological and petrophysical properties to be numerically obtained with confidence since all scales of the pore space can be analyzed and interpreted to calculate properties such as global porosity or relative permeability.
PerGeos Core Profile is a dedicated workspace that enables co-visualization of whole-core CT, core photography, and petrophysical logs in a single environment. Core Profile also creates virtual logs based on whole-core CT data for further calibration and communication of how changes in rock type impact how petrophysical properties are measured down-hole.
The PerGeos Software Petrophysics extension provides a powerful suite of advanced models for rock property analysis and simulations to obtain properties such as absolute permeability, formation factor, thermal properties, molecular diffusion, and MICP porosity.
PerGeos Software provides advanced capabilities for two-phase flow simulation. Properties such as capillary pressure, relative permeability resistivity index, or residual oil saturation can be obtained using this powerful, industry-validated technology.
The Pore Network Statistics extension provides a workflow to generate and extract a pore network model (PNM) based on 3D image data. It provides insight into pore and throat size distributions as a natural outcome of producing a pore network model.
Dual Energy Computed Tomography (DECT)
DECT technology adds value to whole-core CT analysis workflows, by providing you with actual density information that can be compared to density logs and used for further characterization of rock formations. This enables you to compare whole-core CT images across multiple formations and wells.
PerGeos Core Profile is a dedicated workspace that enables co-visualization of whole-core CT, core photography, and petrophysical logs in a single environment. Core Profile also creates virtual logs based on whole-core CT data for further calibration and communication of how changes in rock type impact how petrophysical properties are measured down-hole.
The PerGeos Software Petrophysics extension provides a powerful suite of advanced models for rock property analysis and simulations to obtain properties such as absolute permeability, formation factor, thermal properties, molecular diffusion, and MICP porosity.
PerGeos Software provides advanced capabilities for two-phase flow simulation. Properties such as capillary pressure, relative permeability resistivity index, or residual oil saturation can be obtained using this powerful, industry-validated technology.
The Pore Network Statistics extension provides a workflow to generate and extract a pore network model (PNM) based on 3D image data. It provides insight into pore and throat size distributions as a natural outcome of producing a pore network model.
DECT technology adds value to whole-core CT analysis workflows, by providing you with actual density information that can be compared to density logs and used for further characterization of rock formations. This enables you to compare whole-core CT images across multiple formations and wells.
Shorten your learning curve and maximize your investment with this introductory training specifically designed for new users of Amira, Avizo and PerGeos Software.
The course consists of a lecture with hands-on sessions. The training material highlights the basic features and functionalities of Amira, Avizo and PerGeos Software.
Advanced training
Maximize your investment and reduce your time-to-results with this advanced training specifically designed for existing users of Amira, Avizo and PerGeos Software.
The course consists of a lecture with hands-on sessions. The training material highlights advanced features and functionalities of Amira, Avizo and PerGeos Software.
Custom development
With over 25 years of experience in 3D and image processing and hundreds of custom projects delivered to organizations small and large, Thermo Fisher Scientific can provide you with a solution tailored to fit your specific needs.
We can customize and expand our software solutions at various levels.