Maintenance of LNCaP cells before spheroid generation
After thawing from liquid nitrogen, cells were maintained in Nunclon Delta T25 cell culture flasks in Gibco RPMI medium supplemented with 10% FBS and 1% Pen-Strep for 1 passage before seeding for spheroid generation. ATCC protocol was followed for subculturing.
Materials required
- Nunclon Sphera 96-well plate
- Complete medium (Gibco RPMI medium supplemented with 10% FBS and 1% Pen-Strep)
- 1X PBS
- TrypLE reagent
- Centrifuge with a swinging bucket rotor and holder for 96-well plates
- Countess II Cell Counter or hemocytometer
- 15/50 ml centrifuge tubes
- Multi-channel pipette
- Sterile reagent reservoirs
Protocol for spheroid generation
- Once the flask was 70–80% confluent, medium from the flask was aspirated, the cells were washed once in 1X PBS and dissociated using 1–1.5 ml of TrypLE reagent.
- The TrypLE reagent was neutralized using 4 volumes of complete medium, and live-cell count and viability were captured using Countess II cell counting chamber. Cells with >90% viability were taken for spheroid generation.
- The stock of cells was diluted 1:10 to 1:20 in complete medium to make calculations for cell seeding density easier.
- Seeding cell number was calculated using the cell seeding calculator.
Day 0
- Required number of cells was seeded in respective wells of the Nunclon Sphera plate using a multi-channel pipette. The final volume was maintained at 200 μl.
- The plate was centrifuged at 1,500 rpm for 10 minutes and placed in the incubator at 37°C and 5% CO2.
Media change
- Media was changed 1:1 every alternate day (aspirate 100 μl of spent medium and add 100 μl of fresh medium). The plate was centrifuged at 1,200 rpm for 5 minutes after media change and placed back in the incubator with above mentioned conditions.
Day 4–8
- Spheroids were ready, depending on seeding number.
Skip to Cell viability assay protocol or LIVE/DEAD visualization protocol
Cell Seeding Calculator
(as determined by the Countess Automated Cell Counter)
Number of cells to seed per well
(user-specified number)
... µL
Volume of cell suspension that containsthe specified number of cells
Tips
- Fill the outermost wells of the plate with PBS to prevent evaporation of media during incubation.
- For LNCaP cells, seeding density of >5,000 cells does not result in the formation of good spheroids. At higher density cells tend to fold upon each other, leading to a deformed shape.
- Be gentle while pipetting and ensure the tip is not touching the Sphera 96-well plate surface.
- We have observed that centrifuging the plate after seeding cells facilitates cell aggregation leading to uniform spheroid formation consistently. However, this step is optional.
Morphology of LNCaP spheroids
150–10,000 LNCaP cells were seeded for spheroid generation and brightfield images of spheroids at Day 2, Day 4, Day 6, and Day 8 were captured using EVOS M7000 microscope under 4x magnification. Scale bar = 650 µm.
Characterization of LNCaP spheroids
Assessing cell viability using PrestoBlue-HS cell viability reagent
- On Day 4, cell viability assay was performed using the PrestoBlue HS cell viability reagent.
- The PrestoBlue HS reagent was warmed up to room temperature. Then, 20 μl was added to each well containing 200 μl of medium using a multichannel pipette, and gently mixed by pipetting 2-3 times. Wells containing only fresh medium and the reagent were used as normalization control (blank).
- The plate was incubated at 37°C for 4 hours.
- Following this, fluorescence was read using the Varioskan LUX Multimode Microplate Reader using the following settings:
- Excitation: 560 nm; Emission: 590 nm
- 12 nm excitation bandwidth
- Measurement time: 100 ms
- Bottom optics reading
- Instrument temperature: 37°C
The data was exported to Microsoft Excel for analysis. Individual fluorescence values were normalized to Blank and mean of at least 6 replicates per cell seeding number were plotted using graphing and statistics software. The experiment was repeated 3 times.
Graph showing relative viability of LNCaP spheroids of Day 4 post cell seeding. Error bar represents SEM.
Visualizing live and dead cells using LIVE/DEAD Viability/Cytotoxicity kit
- The population of live and dead cells on Day 4 LNCaP spheroids were visualized by staining them using the LIVE/DEAD Viability/Cytotoxicity kit.
- Working solution was prepared by adding Calcein AM and EthD-1 at a final concentration of 1.5 μM and 3 μM respectively respectively in fresh medium (See our application note for more information on fluorescence staining of spheroids).
- NucBlue Live ReadyProbes Reagent was added (2 drops per ml) to the working solution for nuclear staining of the spheroids. Spent medium of spheroids was changed 1:1 with working solution and incubated at 37°C for 3 hours.
- Following this, they were washed three times with PBS + 5% FBS (1:1 change), and finally resuspended in PBS + 0.5% FBS (1:1 change), each time centrifuging the plate at 500 rpm for 5 minutes at room temperature to settle the spheroids and minimize background during fluorescence imaging.
- The spheroids were then imaged using CellInsight CX7 high-content screening platform under 4X objective. Growing the spheroids for a longer period results in cell death. As shown in the figure, Day 7 spheroids showed reduction in cell viability (red staining).
Graph showing percent of dead cells in LNCaP spheroids on Day 4. Fluorescence reading was captured using Varioskan LUX Multimode Microplate Reader. Values were plotted using GraphPad Prism. Spheroids treated with 70% methanol overnight were used as positive control for dead cells for respective seeding densities.
For Research Use Only. Not for use in diagnostic procedures.