siRNA Details
Gene Symbol: MCM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281520.1 | NP_001268449.1 | 7 | 1114 | ||
NM_001281521.1 | NP_001268450.1 | 7 | 1060 | |||
NM_001281522.1 | NP_001268451.1 | 7 | 1060 | |||
NM_032485.5 | NP_115874.3 | 7 | 1060 | |||
NM_182802.2 | NP_877954.1 | 7 | 1114 | |||
XM_011529387.2 | XP_011527689.1 | 7 | 1023 | |||
XM_017028105.1 | XP_016883594.1 | 7 | 1089 | |||
XM_017028106.1 | XP_016883595.1 | 7 | 1026 | |||
XM_017028107.1 | XP_016883596.1 | 1 | 1385 | |||
GenBank mRNA | AJ439063.1 | - | 7 | 1060 | ||
AK027644.1 | - | 1 | 99 | |||
AK314654.1 | - | 7 | 742 | |||
AY158211.1 | - | 7 | 813 | |||
BC005170.1 | - | 7 | 813 | |||
BC008830.2 | - | 4 | 370 | |||
BC080656.1 | - | 7 | 765 | 4841271 | ||
BC101054.1 | - | 7 | 861 | 40008094 | ||
BC101055.1 | - | 7 | 861 | 40008095 | ||
BC101056.1 | - | 7 | 861 | 40008096 | ||
BC101057.1 | - | 7 | 915 | 40008097 |