siRNA Details
Gene Symbol: CCDC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025004.2 | NP_079280.2 | 5 | 820 | ||
XM_017018346.1 | XP_016873835.1 | 5 | 820 | |||
XM_017018347.1 | XP_016873836.1 | 5 | 820 | |||
XM_017018348.1 | XP_016873837.1 | 5 | 820 | |||
XM_017018349.1 | XP_016873838.1 | 4 | 1961 | |||
XM_017018350.1 | XP_016873839.1 | 5 | 820 | |||
XM_017018351.1 | XP_016873840.1 | 5 | 820 | |||
XM_017018352.1 | XP_016873841.1 | 4 | 470 | |||
XM_017018353.1 | XP_016873842.1 | 5 | 820 | |||
GenBank mRNA | AK023277.1 | - | 5 | 820 | ||
BC018540.1 | - | 5 | 734 |