siRNA Details
Gene Symbol: SLC17A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020309.3 | NP_064705.1 | 7 | 1015 | ||
GenBank mRNA | AB032436.1 | - | 7 | 864 | ||
AK091061.1 | - | 3 | 266 | |||
AK095659.2 | - | 6 | 2294 | |||
AK289622.1 | - | 4 | 423 | |||
AK294226.1 | - | 7 | 821 | |||
AK294405.1 | - | 7 | 864 | |||
BC059379.1 | - | 7 | 990 | 5262415 |