siRNA Details
Gene Symbol: PHF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278221.2 | NP_001265150.1 | 9 | 1554 | ||
NM_001321126.1 | NP_001308055.1 | 10 | 1630 | |||
NM_001321127.1 | NP_001308056.1 | 10 | 1763 | |||
NM_016483.6 | NP_057567.3 | 10 | 1671 | |||
GenBank mRNA | AF151060.2 | - | 8 | 720 | ||
AK223531.1 | - | 9 | 888 | |||
AK292221.1 | - | 10 | 1477 | |||
AL137671.1 | - | 3 | 1477 | |||
AY014283.2 | - | 10 | 1245 | |||
BC022002.1 | - | 10 | 1530 | 4822315 | ||
BU853281.1 | - | 4 | 389 |