siRNA Details
Gene Symbol: ADH7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000673.4 | NP_000664.2 | 6 | 1072 | ||
NM_001166504.1 | NP_001159976.1 | 6 | 885 | |||
GenBank mRNA | AK223472.1 | - | 6 | 866 | ||
AK301657.1 | - | 6 | 848 | |||
AK301696.1 | - | 6 | 885 | |||
AK310811.1 | - | 5 | 835 | |||
AK312854.1 | - | 6 | 995 | |||
BC131512.1 | - | 6 | 867 | 40083994 | ||
CD014081.1 | - | 7 | 948 | |||
L33179.1 | - | 6 | 795 | |||
S77168.1 | - | 1 | 117 | |||
U07821.1 | - | 6 | 879 | |||
U09623.1 | - | 6 | 813 | |||
X76342.1 | - | 6 | 855 |