siRNA Details
Gene Symbol: Senp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144851.5 | NP_659100.1 | 4 | 489 | ||
GenBank mRNA | AK028565.1 | - | 4 | 231 | ||
AK035581.1 | - | 4 | 325 | |||
AK035723.1 | - | 4 | 593 | |||
AK054521.1 | - | 4 | 490 | |||
AK088597.1 | - | 4 | 233 | |||
AK138013.1 | - | 4 | 290 | |||
BC023129.1 | - | 3 | 203 | 3595534 |