siRNA Details
Gene Symbol: LOC294154 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039607.1 | NP_001034696.1 | 1, 2 | 183 | ||
XM_008772799.1 | XP_008771021.1 | 1, 2 | 377 | |||
GenBank mRNA | BC112320.1 | - | 1, 2 | 183 | 7933524 |