This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MDM2 SIRNA Assays ›
Gene Symbol:
MDM2
Gene Name:
MDM2 proto-oncogene
Gene Aliases:
ACTFS, HDMX, hdm2
Chromosome Location:
Chr.12: 68808149 - 68845544 on Build GRCh38
UniGene ID:
Hs.484551
Species:
Human
siRNA ID
HSS142911
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MDM2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001145337.2
NP_001138809.1
4
257
NM_001145339.2
NP_001138811.1
4
507
NM_002392.5
NP_002383.2
4
507
XM_005268872.4
XP_005268929.1
4
435
XM_006719399.3
XP_006719462.1
2
90
GenBank mRNA
AF092843.1
-
4
215
AF092844.1
-
4
215
AF385323.1
-
3
183
AJ430612.1
-
3
183
AJ430614.1
-
3
183
AJ491698.1
-
3
183
AJ491699.1
-
4
270
AJ491700.1
-
3
183
AK290341.1
-
4
457
AM902642.1
-
4
276
AM902646.1
-
3
183
AM902647.1
-
4
270
AM902650.1
-
3
183
AM902651.1
-
3
183
AM902654.1
-
3
183
AM902655.1
-
3
183
AM902658.1
-
4
270
AM902659.1
-
3
183
AM902660.1
-
3
183
AM902666.1
-
3
183
AM902667.1
-
3
183
AM902668.1
-
3
183
AM902669.1
-
3
183
AM902670.1
-
2
108
AM902672.1
-
3
183
AM902678.1
-
3
183
AM902680.1
-
3
183
AM902681.1
-
3
183
BC152384.1
-
3
186
BC152390.1
-
3
186
BE296905.1
-
4
472
BT007258.1
-
3
183
EU076747.1
-
4
257
EU076749.1
-
4
257
GQ848196.1
-
2
108
HM213901.1
-
3
183
JX015379.1
-
3
183
JX015389.1
-
3
183
JX015390.1
-
3
183
JX015391.1
-
3
183
JX015392.1
-
3
183
M92424.1
-
4
494
U33203.1
-
3
183
Z12020.1
-
4
494
Back To Top
More Information
Panther Classification:
Molecular Function -
chromatin/chromatin-binding, or -regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
blood vessel development
blood vessel remodeling
regulation of heart rate
atrioventricular valve morphogenesis
endocardial cushion morphogenesis
ventricular septum development
atrial septum development
protein complex assembly
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
traversing start control point of mitotic cell cycle
positive regulation of cell proliferation
response to toxic substance
response to carbohydrate
response to iron ion
positive regulation of gene expression
negative regulation of protein processing
viral process
protein ubiquitination
protein sumoylation
peptidyl-lysine modification
protein destabilization
response to magnesium ion
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
protein localization to nucleus
regulation of protein catabolic process
response to cocaine
response to drug
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
response to morphine
negative regulation of DNA damage response, signal transduction by p53 class mediator
establishment of protein localization
response to ether
negative regulation of transcription, DNA-templated
positive regulation of mitotic cell cycle
response to antibiotic
positive regulation of protein export from nucleus
response to steroid hormone
cardiac septum morphogenesis
cellular response to hydrogen peroxide
negative regulation of cell cycle arrest
cellular response to antibiotic
cellular response to vitamin B1
cellular response to alkaloid
cellular response to growth factor stimulus
cellular response to peptide hormone stimulus
cellular response to estrogen stimulus
cellular response to hypoxia
cellular response to UV-C
regulation of signal transduction by p53 class mediator
negative regulation of signal transduction by p53 class mediator
p53 binding
ubiquitin-protein transferase activity
protein binding
zinc ion binding
ligase activity
SUMO transferase activity
enzyme binding
ubiquitin protein ligase binding
identical protein binding
peroxisome proliferator activated receptor binding
ubiquitin protein ligase activity
scaffold protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-7745c474b-b52np:80/100.66.77.222:80.
git-commit: c3c9db9cc82507548abdd0ae6a4ade3f81f66292
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.20.0-2024.11.45-1.0