siRNA Details
Gene Symbol: GPR20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005293.2 | NP_005284.2 | 2 | 680 | ||
GenBank mRNA | AB209066.1 | - | 1 | 338 | ||
BC117409.1 | - | 1 | 684 | 40125960 | ||
JQ308168.1 | - | 1 | 591 |