siRNA Details
Gene Symbol: RUNX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001890.2 | NP_001001890.1 | 2 | 1891 | ||
NM_001122607.1 | NP_001116079.1 | 2 | 1891 | |||
NM_001754.4 | NP_001745.2 | 5 | 584 | |||
XM_005261068.3 | XP_005261125.1 | 3 | 847 | |||
XM_005261069.4 | XP_005261126.1 | 5 | 620 | |||
XM_011529766.2 | XP_011528068.1 | 5 | 695 | |||
XM_011529767.2 | XP_011528069.1 | 4 | 581 | |||
XM_011529768.2 | XP_011528070.1 | 4 | 581 | |||
XM_011529770.2 | XP_011528072.1 | 5 | 620 | |||
XM_017028487.1 | XP_016883976.1 | 3 | 389 | |||
GenBank mRNA | AK226159.1 | - | 4 | 581 | ||
AK310587.1 | - | 5 | 573 | |||
BC110828.1 | - | 4 | 479 | |||
BC136380.1 | - | 5 | 541 | 9020367 | ||
BC136381.1 | - | 5 | 540 | 9020368 | ||
BC144053.1 | - | 5 | 540 | 9052573 | ||
D10570.1 | - | 2 | 1081 | |||
D43967.1 | - | 2 | 1891 | |||
D43968.1 | - | 2 | 1891 | |||
D43969.1 | - | 4 | 838 | |||
D89788.1 | - | 3 | 281 | |||
D89789.1 | - | 3 | 281 | |||
D89790.1 | - | 3 | 281 | |||
L34598.1 | - | 2 | 991 | |||
S60998.1 | - | 2 | 321 | |||
U19601.1 | - | 4 | 394 | |||
X79549.1 | - | 2 | 1881 |