siRNA Details
Gene Symbol: LETM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199659.2 | NP_001186588.1 | 10 | 1484 | ||
NM_001199660.2 | NP_001186589.1 | 10 | 1323 | |||
NM_001286787.1 | NP_001273716.1 | 10 | 1470 | |||
NM_001286819.1 | NP_001273748.1 | 10 | 1555 | |||
NM_144652.3 | NP_653253.1 | 9 | 1340 | |||
XM_006716291.3 | XP_006716354.1 | 10 | 1445 | |||
XM_011544406.1 | XP_011542708.1 | 10 | 1444 | |||
XM_011544407.1 | XP_011542709.1 | 10 | 1406 | |||
XM_011544409.2 | XP_011542711.1 | 10 | 1724 | |||
XM_011544410.2 | XP_011542712.1 | 7 | 972 | |||
XM_017013051.1 | XP_016868540.1 | 10 | 1787 | |||
XM_017013052.1 | XP_016868541.1 | 10 | 1515 | |||
XM_017013053.1 | XP_016868542.1 | 10 | 3861 | |||
XM_017013054.1 | XP_016868543.1 | 10 | 1500 | |||
XM_017013055.1 | XP_016868544.1 | 10 | 1643 | |||
XM_017013056.1 | XP_016868545.1 | 10 | 1489 | |||
XM_017013057.1 | XP_016868546.1 | 9 | 3905 | |||
GenBank mRNA | AK058138.1 | - | 9 | 1279 | ||
AK302317.1 | - | 10 | 1423 | |||
AY173945.1 | - | 10 | 1415 | |||
BC029541.1 | - | 7 | 908 | 5297908 | ||
BC069830.1 | - | 10 | 1404 | |||
CB242043.1 | - | 2 | 367 |