siRNA Details
Gene Symbol: POLG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126131.1 | NP_001119603.1 | 21 | 3663 | ||
NM_002693.2 | NP_002684.1 | 21 | 3675 | |||
GenBank mRNA | AK130624.1 | - | 4 | 549 | ||
AK291281.1 | - | 21 | 3694 | |||
BC042571.1 | - | 21 | 3660 | 4384475 | ||
BC050559.1 | - | 21 | 3644 | 6042595 | ||
D84103.1 | - | 20 | 3499 | |||
U60325.1 | - | 20 | 3552 | |||
X98093.1 | - | 21 | 3675 |