siRNA Details
Gene Symbol: Chek1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007691.5 | NP_031717.2 | 3 | 345 | ||
GenBank mRNA | AF016583.1 | - | 2 | 254 | ||
AK011258.1 | - | 3 | 289 | |||
AK033179.1 | - | 3 | 315 | |||
AK045336.1 | - | 3 | 287 | |||
AK166675.1 | - | 3 | 346 | |||
BC053394.1 | - | 3 | 281 | |||
BC064445.1 | - | 2 | 745 | |||
BC100386.1 | - | 2 | 420 | 30481390 |