siRNA Details
Gene Symbol: CCDC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025004.2 | NP_079280.2 | 16 | 3373 |
Gene Symbol: CCDC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025004.2 | NP_079280.2 | 16 | 3373 |