siRNA Details
Gene Symbol: MCHR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005297.3 | NP_005288.3 | 2 | 2103 | ||
GenBank mRNA | AF490537.2 | - | 2 | 2186 |
More Information
Set Membership: |
G-protein coupled receptor |