siRNA Details
Gene Symbol: Slc36a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153170.3 | NP_694810.2 | 10 | 2093 | ||
GenBank mRNA | AF453744.1 | - | 10 | 2044 | ||
AF512429.1 | - | 10 | 2116 | |||
AK041041.1 | - | 10 | 2085 | |||
AK079053.1 | - | 10 | 2058 | |||
AK156409.1 | - | 10 | 2092 | |||
BC044800.1 | - | 10 | 2091 | 4189221 |