siRNA Details
Gene Symbol: DCUN1D3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173475.3 | NP_775746.1 | 2 | 571 | ||
GenBank mRNA | AK123719.1 | - | 2 | 502 | ||
AY364247.1 | - | 1 | 311 | |||
BC040442.1 | - | 2 | 551 | 5590225 | ||
BX648998.1 | - | 2 | 453 |