siRNA Details
Gene Symbol: CSNK1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001893.4 | NP_001884.2 | 5 | 1025 | ||
NM_139062.2 | NP_620693.1 | 5 | 1025 | |||
NR_110578.1 | 4 | 820 | ||||
XM_005256336.3 | XP_005256393.1 | 5 | 1018 | |||
XM_005256337.4 | XP_005256394.1 | 5 | 1018 | |||
XM_017024199.1 | XP_016879688.1 | 5 | 782 | |||
GenBank mRNA | AB091044.1 | - | 5 | 737 | ||
AB209463.1 | - | 4 | 526 | |||
AK128863.1 | - | 2066 | ||||
AK291758.1 | - | 5 | 1007 | |||
AK295638.1 | - | 4 | 576 | |||
AK296969.1 | - | 4 | 509 | |||
AK303365.1 | - | 6 | 758 | |||
AK308852.1 | - | 1 | 18 | |||
BC003558.1 | - | 5 | 996 | 3546633 | ||
BC015775.1 | - | 5 | 1004 | 4849869 | ||
U29171.1 | - | 5 | 865 |