siRNA Details
Gene Symbol: FADS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013402.4 | NP_037534.3 | 12 | 2867 | ||
XM_011545022.2 | XP_011543324.1 | 12 | 2541 | |||
GenBank mRNA | AF035284.1 | - | 1 | 813 | ||
AF084558.1 | - | 12 | 2652 | |||
AL512760.1 | - | 12 | 2648 | |||
AL834479.1 | - | 4 | 1511 |