The SOLiD® EZ Bead™ System provides an automated solution for reproducible templated bead preparation in less than 1 hour of hands-on time. The system modularity and quality control checkpoints provide greater experimental control. Additional support for multiplexed libraries further maximizes time and cost savings.
- Automated - template bead preparation in less than 1 hour of hands-on time
- Modular - designed to fit individual throughput and laboratory needs, the SOLiD® EZ Bead™ System’s three modules, the SOLiD® EZ Bead™ Emulsifier, the SOLiD® EZ Bead™ Amplifier, and the SOLiD™ EZ Bead™ Enricher, facilitate customizable instrument configurations to align with your research
- Scalable - supports bead outputs for 1-, 4-, and 8-well slide configurations with corresponding E10, E20, and E80 prepackaged reagents to accommodate a variety of high-, medium-, and low-throughput applications.
Order:
Product Information
Video
For Research Use Only. Not for use in diagnostic procedures.