dataTaker loggers have been designed to provide logging capability for environmental monitoring systems. The loggers support a wide range of weather sensors, air quality sensors, soil sensors, and other sensors within environmental area from various manufacturers.
With SDI12 support, low power consumption, various communication options, and high capacity memory, dataTaker loggers are ready to be deployed on remote locations for long term monitoring.
DT90 | DT82E series | DT80 series | DT85 series |
---|---|---|---|
Low cost battery powered data logger, for remote sites | Low cost, low power data logger | Intermediate data logger | Advanced data logger |
IP67 | IP54 | ||
Integrated 4G modem | - | Options for integrated modem | |
Up 8 analog inputs, dependent on model | 2 analog input channels | 5 analog input channels, expandable to 300 | 16 analog input channels, expandable to 960 |
3 digital input/counter channels | 4 digital input/counter channels | 8 digital input/counter channels | |
~500,000 data points | ~10,000,000 data points | ||
USB Port | - | USB port | |
- | 1 serial ‘smart sensor’ ports | 2 serial ‘smart sensor’ ports | |
- | Web and FTP client, RS232/485 | ||
SDI-12, Modbus, USB memory for easy data and program transfer |
Environmental logging accessories
Increasing the channel capacity of the dataTaker DT80 and DT85 range is made very easy by adding dataTaker Channel Expansion Modules (CEM20). Each CEM20 connects 20 universal data logging channels to the dataTaker data logger.
dataTaker enclosures give you the flexibility to install your dataTaker system in a harsh or otherwise unsuitable environment by providing protection against external conditions such as water, dust, and accidental damage. These enclosures are suitable for all dataTaker systems and can be used for, industrial, weatherproof, or portable applications.
Resources
Brochures: