Find valuable information.
Optimize your experiments to get the best results. We’ve compiled a detailed knowledge base of the top tips and tricks to meet your analyses needs.
View the relevant questions below:
Having problems with your experiment? Visit
Getting Started
ProSightPC is a standalone software supporting database creation, analysis of infusion data, and high-throughput LC/MS data. ProSightPD is a collection of nodes enabling top-down LC/MS data to be searched in the Proteome Discoverer application.
For ProSight PC:
- Microsoft™ Windows™ 7 x64 Professional (English version) with current service pack
- ProSight Lite (freeware from prosightlite.northwestern.edu)
For ProSight PD:
- ProSightPC
- Proteome Discoverer (x64 version)
- Thermo Proteome Discoverer ThirdParty Components installer (also installs the ProSightPD nodes and ProSightPD with High Mass nodes)
- Microsoft™ Windows™ 7 x64 Professional (English version) with current service pack
- ProSight Lite (freeware from prosightlite.northwestern.edu)
RAW files can be read directly into either ProSightPC or ProSightPD (via Proteome Discoverer) for detailed descriptions of building and executing data analysis workflows in both software please refer to the Quick Start Guides which can be found below.
Databases with PTMs and FASTA files (no PTMs) can be found here: proteinaceous.net/database-warehouse/
Click the Add Experiment icon, or choose Tools > Experiment Adder to open the Experiment Adder dialog box
In the Fragmentation Methods area, select the fragmentation method.
In the Precursor Ion Data/Type box, select the precursor mass type, either Manual or Upload. If you select Manual in the Precursor Ion Data/Type list, select the mass type of the precursor ion in the Mass Type box (either Monoisotopic or Average) and enter the precursor mass. If you select Upload in the Precursor Ion Data/Type list, enter the path and name of the ASCII text file or files containing the precursor ion data in the Text File box, or click Browse to browse for them.
(Optional) Select a predefined search in the box near the bottom of the dialog box.
Need more information? Contact us ›
For Research Use Only. Not for use in diagnostic procedures.