Applied Biosystems AmpliTaq DNA Polymerase is a recombinant preparation of Taq DNA Polymerase. AmpliTaq DNA Polymerase is used in standard research applications such as routine PCR, genotyping, and colony PCR.
AmpliTaq DNA Polymerase is available in different formulations as summarized in the table below. AmpliTaq 360 DNA Polymerase and AmpliTaq LD DNA Polymerase are ultra-pure preparations that are used for experiments where it is important to reduce the incidence of false positives.
AmpliTaq DNA Polymerase product formats
AmpliTaq DNA polymerase | AmpliTaq 360 DNA polymerase | |
---|---|---|
Amplicon size | Up to 5 kb | Up to 5 kb |
Product overhang | 3′ A | 3′ A |
Exonuclease activity | 5′→ 3′ | 5′→ 3′ |
GC-rich amplification | No | Yes |
Stand-alone enzyme | ||
Stand-alone enzyme from low DNA (LD) preparation1 |
1 Applied Biosystems AmpliTaq DNA Polymerase, LD (Low DNA), is the same enzyme as AmpliTaq DNA Polymerase, but is further purified through a proprietary separation process to ensure that residual bacterial DNA sequences are substantially reduced. The enzyme is quality-control tested to verify that fewer than 10 copies of bacterial 16S ribosomal RNA gene sequences are present in a standard 2.5 unit aliquot.
Note: If you are looking for the hot-start version of AmpliTaq DNA Polymerase, please see AmpliTaq Gold DNA Polymerase.
AmpliTaq 360 Polymerase reagents amplify a broad range of targets
Figure 1. AmpliTaq 360 DNA Polymerase Amplifies a Broad Range of Targets. The gel image shows data produced with AmpliTaq 360 DNA Polymerase. PCR reactions were performed using 1 ng of template DNA and 1.25 units of enzyme in each 50 µL reaction. Annealing was uniform across the selected targets. Amplicons ranged from 300 to 1,400 base pairs (bp) in length with an average length of 553 bp. Each reaction was performed in duplicate. Amplicons are labeled as follows: High AT, Easy Amplification, Primer Dimer, Homopolymer, Sequencing Challenge, Long, and High GC. The high GC target reactions included 2–10 µL of 360 GC Enhancer depending upon the target used.
Figure 2. Sensitivity of AmpliTaq 360 DNA Polymerase for Detection of Low Copy Targets. The gel image shows the amplification products of the beta-actin gene performed with 0 to 80 copies of input DNA, using 2.0 mM MgCl2 and standard thermal cycling conditions with a 30 second extension time.
Figure 1. AmpliTaq 360 DNA Polymerase Amplifies a Broad Range of Targets. The gel image shows data produced with AmpliTaq 360 DNA Polymerase. PCR reactions were performed using 1 ng of template DNA and 1.25 units of enzyme in each 50 µL reaction. Annealing was uniform across the selected targets. Amplicons ranged from 300 to 1,400 base pairs (bp) in length with an average length of 553 bp. Each reaction was performed in duplicate. Amplicons are labeled as follows: High AT, Easy Amplification, Primer Dimer, Homopolymer, Sequencing Challenge, Long, and High GC. The high GC target reactions included 2–10 µL of 360 GC Enhancer depending upon the target used.
Figure 2. Sensitivity of AmpliTaq 360 DNA Polymerase for Detection of Low Copy Targets. The gel image shows the amplification products of the beta-actin gene performed with 0 to 80 copies of input DNA, using 2.0 mM MgCl2 and standard thermal cycling conditions with a 30 second extension time.
Resources
Product manuals
White paper and fact sheet
PCR education
Learn about the history of PCR, setup considerations, importance of DNA polymerase choice, common methods and applications, and troubleshooting.
Molecular biology webinars
View our webinar series to learn more about molecular biology topics such as reverse transcription, PCR and cloning.
Support
PCR and cDNA synthesis support center
Find tips, troubleshooting help, and resources for your end-point PCR and cDNA applications.
OEM & customized PCR solutions
Discover customizable manufacturing solutions, product labeling, and packaging capabilities for your specific requirements.
Contact us
Email or call our Technical Application Scientists for additional questions regarding PCR enzymes and master mixes.
For Research Use Only. Not for use in diagnostic procedures.