">
Explore our water purification systems virtually to determine which is best for your lab
Our 3D product tours enable you to view and rotate, zoom, discover and interact with each model. Embedded in each tour are detailed descriptions of key features and capabilities.
Aquanex Ultrapure Water Purification System
The Thermo Scientific Aquanex system is designed to truly help make life easier in lab with an intuitive touch screen interface, easy-access consumables, and smart dispensing features.  The easy-to-use interface caters to users that need secured access and seamless event logging, system monitoring, and reporting.  A new innovative cartridge design provides low TOC, a longer cartridge life, and an easy-to-change design.
Smart2Pure Pro Water Purification System
The Smart2Pure Pro system is a compact system which converts tap water to into ASTM Type 1. The Smart2Pure Pro UV/UF 16 LPH offers the choice of a 30 L or 60 L reservoir for customizable storage capacity and ability to dispense both ASTM Type 1 and Type II water.
">