The MAGnify™ ChIP system uses a novel magnet that is compatible with 0.2 mL PCR strip tubes. The new
DynaMag-PCR magnet is optimized for efficient magnetic separation of small sample volumes using DynaBeads.This magnet is critical for enabling the use of reduced starting cell numbers and volumes for immunoprecipitation and washing steps. This novel magnetic separator allows you to achieve results quickly and easily while offering the freedom to simultaneously perform multiple ChIP assays in PCR tubes with a multi-channel pipettor.
- Optimal working volume: Up to 200 µl
- Holds up to 16 0.2 ml PCR tubes (individual or strip tubes)