Achieve uniform coating excellence
We offer a complete line of non-contact metallic and non-metallic coating weight measurement systems that provide precise, real-time measurements that meet the tightest specifications of any application while maximizing raw material use. From hot-dipped or electro-deposited to roll coated or sprayed-on coatings, you can rely on us for accurate coating weight measurements that help assure uniform coverage for the highest quality finished products.
Featured coating weight gauging products
Thermo Scientific paint thickness gauge provides fast, accurate and reliable profile measurement of paint and organic coatings, providing essential information to optimize your production process.
Thermo Scientific oil thickness gauge provides fast, accurate, non-contact coating weight measurement of oil coatings applied to flat sheet galvanneal products. It is especially suited for thin and ultra-thin coatings and for selective measurement of multilayer coatings.
Thermo Scientific metallic coating gauge provides fast, accurate, non-contact measurement of coatings applied to flat sheet galvanneal products, helping to ensure coating uniformity and product quality.
Metals gauging processes
Video: Metals coating weight & thickness measurement gauges
Optimize your steel manufacturing process
Learn why Thermo Scientific metals thickness and coating weight measurement products make every stage of the steel-making process more efficient.