">
How do I know what format is right for my laboratory?
Our real-time PCR platform supports multiple format options to help research laboratories operate efficiently through fluctuating seasonal monthly and weekly sample volume needs.
How many samples are you expecting to run? Consider your daily and weekly testing volume and throughput needs to find the right solution for your laboratory.
  Single tube assays Plates
Cards
OpenArray plates
Format Individual tubes or assay tube sets in a variety of reaction sizes 96-and 384-well plates with assays pre-spotted into the wells 384-well microfluidic cards with assays pre-spotted 3,000 through-holes with assays pre-spotted
Assay type Singleplex or Spectral Multiplex Singleplex or Spectral Multiplex Singleplex Spatial-Multiplex
Typical # of targets 1 to 5 4 to 24 24 or 48 18 or 56
Use case For smaller panels in any volume (1 to 1000s) For mid-size panels in mid-throughput (10–50) For large panels in low throughput (10s) For mid to large panels in high throughput (48–1000s)
Your advantage High flexibility for low throughput Supports automation for medium throughput Convenience—no robotics needed Higher throughput for routine applications or large projects 
Confidently design a TrueMark custom qPCR research panel with TaqMan assays
Whether you want to modify from an existing design or start with your own target list, we’re here to walk you through the process.
Format Approximate turnaround time
(after order has been placed)
Single tubes 5 days
Cards or plates 3–5 weeks
OpenArray plates 5 weeks
Complete this form to begin your design process. A specialist will reach out for a one-on-one consultation to help get you what you need.
Utilizing the Microbial Search Tool to browse our assay catalogue
First, visit the microbial search tool. Then:
  1. Use the search field to find your target(s) of interest 
  2. Filter based on categories: Organism, Species/Strain, Assay Attributes, or Assay Plex
  3. Option to search for multiple targets
Below is a selection of how-to videos to help you navigate the microbial search tool. You’ll be a pro in no time. But if you do require additional assistance with assay selection, please complete this form for help with panel design and ordering.
For single-target searches:
Pathogen detection assays
In this video, learn how to search for pathogen detection assays in the microbial search tool.
Antibiotic resistance assays
In this video, learn how to search for antibiotic resistance assays in the microbial search tool.
Performing a batch search:
In this video, learn how to perform a batch search for pathogen detection assays in the microbial search tool.
Configuring an OpenArray panel
In this video, learn how to configure an OpenArray panel that includes both pathogen detection and antibiotic resistance assays through the microbial search tool.
">