Thermo Scientific Surface Protectors can help make your lab a safer place.
Thermo Scientific Versi-Dry bench protectors quickly absorb and contain spills and cushion breakable labware. Use Versi-Dry material as a bench protector, shelf liner, cabinet liner or for wiping up isolated spills. The skid-resistant polyethylene backing protects your benchtop and other lab surfaces from damage and contamination caused by liquid drips and spills. The cloth-like top layer is long lasting and will not fall apart, even when wet. Thermo Scientific Nalgene CleanSheets are made of 0.125" (3.2mm) thick closed-cell polyethylene foam and can be used as a bench, drawer or sink liner or as a titration background. CleanSheets provide cushioning and non-absorbent liquid barrier protection.
Which surface protector is best for you?
Versi-Dry Surface Protectors | Super Versi-Dry Surface Protectors | CleanSheets Surface Protectors |
Absorbs 750mL of liquid/m2 | Absorbs 1,050mL of liquid/m2 | Non-absorbent |
Mat sizes: 46 x 51cm; 18 x 20 in. | Mat sizes: 46 x 51cm; 18 x 20 in. 46 x 102cm; 18 x 40 in. 51 x 110cm; 20 x 43 in. | Mat sizes: not available in mats |
Roll sizes:
51cm x 30m; 20 in. x 100 ft. 51cm x 46m; 20 in. x 150 ft. 51cm x 91m; 20 in. x 300 ft. |
Roll sizes:
51cm x 30m; 20 in. x 100 ft. 51cm x 76m; 20 in. x 250 ft. 102cm x 61m; 40 in. x 200 ft. | Roll sizes: 31cm x 15.2m; 12 in. x 50 ft. 46cm x 15.2m; 18 in. x 50 ft. |
White fibrous top layer | Blue fibrous top layer | NA |
Polyethylene backer | Polyethylene backer | White 0.125" (3.2mm) thick closed-cell polyethylene foam |
Thermo Scientific surface protectors offer protection from the following chemical spills:
Acetone
Hydrofluoric Acid, 10%* Potassium Hydroxide, 30%* Ammonium Hydroxide, 30%* Methylene Chloride Sulfuric Acid, 10% Chlorine Bleach |
Methyl Ethyl Ketone (MEK)
Sodium Hydroxide, 10N* Chlorobenzene Methyl Iso Butyl Ketone (MIBK) Toluene Chloroform |
Mineral Spirits
Xylene Ethanolamine Nitric Acid, 10%* Hydrochloric Acid, 10%* Phosphoric Acid, 10%* |
*Highest concentration tested
Resources
Certificate of Compliance
Receive a Certificate of Compliance and Certificate(s) of Irradiation (if applicable) on purchased Thermo Scientific Nalgene and Nunc products.
Support
For a more detailed chemical resistance guide by chemical and application temperature, please refer to the Nalgene Chemical Resistance Wall Chart or contact technicalsupport@thermofisher.com.