">
Advancing Veterinary Care
Thermo Scientific Indiko and Indiko Plus analyzers for vet clinics and labs
On this page:
Fully-automated, random access analyzer for fast and reliable companion animal care
Thermo Scientific Indiko and Indiko Plus clinical chemistry analyzers are now available for use in mid-sized veterinary clinics and labs. These fully-automated, random access bench top analyzers offer efficiency and high-quality results, providing veterinary professionals with tools to enhance pet sample analysis.
Analyze a greater number of samples in a shorter time and give your business an opportunity to grow!
This webinar aims to equip veterinary professionals with the tools and knowledge necessary to deliver optimal care and improve the overall well-being of our animal companions.
Our animal companions deserve high-quality care and testing – choose the Indiko Vet Solution!
Equip your clinic with the Indiko clinical chemistry analyzer. The Indiko analyzers provide veterinarians with adaptable solutions to efficiently process laboratory tests on site—and in any order—with random access capability.
Extensive automation: easy to use with rapid results
Keep the day on track
  • Analyze samples in any order with random-access capability
  • Continuous access to samples, reagents, and cuvettes ensures uninterrupted test runs
  • User-definable tests can be easily created for clinical chemistry and electrolyte assays
More flexibility
  • Simultaneously run multiple tests with up to 108 samples and 42 reagents
  • Make the most of each sample with individual tests or choose from predefined profiles, including STAT requests to prioritize results
  • Reduce cost and waste, with kit sizes optimized for small and medium-sized labs
More time
  • Simplify laboratory operations with the load-and-go feature
  • Barcoded reagents, real-time monitoring and automatic flagging of abnormal values streamline result analysis
  • Ensure maximal uptime with minimal required maintenance
Rapid, reliable results
  • Access rapid results within minutes for timely insights and effective care decisions
  • Reduce errors and manual work with automated operations
  • Disposable cuvettes ensure accuracy and speed
  • Secure and traceable data handling ensures data integrity
Clinical chemistry reagents
Ion selective electrodes
Vet Alkaline Phosphatase (IFCC) Plus
Vet CK (IFCC)
Vet Lipase
Vet Potassium
Vet ALT/GPT (IFCC)
Vet Creatinine (Enzymatic)
Vet Magnesium
Vet Sodium
Vet Amylase (IFCC)
Vet Creatinine (Jaffe)
Vet Phosphorus
Vet Chloride
Vet AST/GOT (IFCC)
Vet Gamma-GT (IFCC)
Vet Pyridoxal Phosphate
Vet Bicarbonate
Vet Glucose (GOD-POD)
Vet Total Protein Plus
Vet Bilirubin direct
Vet Glucose (HK)
Vet Triglycerides
Vet Bilirubin total (DCA)
Vet HDL-Cholesterol
Vet U/CSF Protein
Vet Calcium
Vet LDL-Cholesterol
Vet Urea
Vet Cholesterol
Vet Iron
Vet Uric Acid (AOX)
Vet Cholinesterase
Vet LDH (IFCC)
Choose the analyzer that is right for you
Indiko system
Indiko clinical chemistry analyzer
Indiko Plus clinical chemistry analyzer
Capacity Photometric tests/hour: up to 200
ISE tests/hour: up to 120
Photometric tests/hour: up to 350
ISE tests/hour: up to 135
On-board sample capacity Max 90 with 5 x 18-pos. sample rack Max 108 with 6 x 18-pos. sample rack
On-board reagent capacity Max 30 with 1 x 9- or 18-pos. sample rack and 5 x 6-pos. reagent rack Max 42 reagent positions
Walk-away time Up to 2 hours Up to 3 hours
Water consumption 1.5 liters/hour 2.5 liters/hour
Dimensions (WxDxH) & weight
75 cm x 70 cm x 62 cm / 130 cm with the open cover & 85 kg
29.5 in x 27.6 in x 24.4 in / 51 in with the open cover & 187 lbs
94 cm x 70 cm x 62 cm / 130 cm with the open cover & 110 kg
37 in x 27.6 in x 24.4 in / 51 in with the open cover & 242 lbs
Want to learn more?
Contact a specialist today to learn how Indiko systems can simplify and speed up your lab work. 
Test consistently and confidently with the largest offering of high-quality animal health molecular solutions
When it comes to preserving the health of an entire herd, your customers' decisions, reputations, and livelihoods are dependent upon the diagnostic test results your laboratory provides. That’s why we’ve dedicated ourselves to creating the most dynamic and diverse portfolio of solutions to help ensure that you’re providing quick, accurate, and actionable results.
">