Specifications
TypeClinical Chemistry Analyzer
DescriptionIndiko without ISE Unit
CapacityOn to board capacity: Combined sample and reagent disk with 9 to or 18 to position sample racks and 6 to position reagent racks. Max 6 racks.
Cuvettes9 x 10-position single-use cuvettes, totally 90 cuvette positions
Distilled Water Consumption1.5 L/hr.
Certifications/ComplianceCE, FCC, UL, CAN/CSA
ReagentsReagent volume: 2 - 240uL (1 to 4 reagent additions/test)
Depth (English)27.6 in.
Depth (Metric)70 cm
Height (English)24.4 in.
Height (Metric)62 cm
Width (English)29.5 in.
Width (Metric)75 cm
Frequency50/60 Hz +/-5%
Voltage100/240 V +/-10%
Weight (Metric)85 kg
Unit SizeEach