Innovations in clinical research and toxicology for LC-MS assays
Transitioning to a new platform or technologies, while challenging, can lead to faster results, easier sample preparation, and more profitability. Thermo Scientific products use cutting-edge technologies that enable you to develop reliable and robust clinical research assays.
On this page
In order to efficiently utilize the mass spectrometer and eliminate downtime, the Thermo Scientific Transcend TLX-1 TurboFlow and Transcend LX-2 systems give the user the ability to use two independent liquid chromatography channels to analyze your clinical assay. Both platforms are available in 1- ,2-, and 4-channel configurations.
Thermo Scientific Transcend Duo UHPLC systems provide two independent channels with Vanquish pump technology to improve your productivity by saving time, reducing cost per sample, increasing capacity without added bench space, and using resources more efficiently.
Technical notes
Webinar
eBook
Posters
Case Studies
The Thermo Scientific VeriSpray PaperSpray Ion Source employs paper spray technology, a direct ionization technique that improves throughput, reduces cost per test and minimizes sample preparation. The technology is well suited for TDM, toxicology, and clinical research applications where time and cost per sample is a concern. The VeriSpray PaperSpray ion source uses simplified workflows that eliminate the need for more complex analytical techniques and enable the collection of quantitative and qualitative data by mass spectrometry.
Technical notes
Additional Ion Source resources
High-field asymmetric waveform ion mobility spectrometry Thermo Scientific (FAIMS) Pro Duo interface uses ion characteristics such as: charge state, shape, conformation, and size of the gas phase ion to reduce chemical noise and matrix interferences. These attributes provide assays with improved robustness and increased sensitivity.
Technical notes
Specification sheet
In order to efficiently utilize the mass spectrometer and eliminate downtime, the Thermo Scientific Transcend TLX-1 TurboFlow and Transcend LX-2 systems give the user the ability to use two independent liquid chromatography channels to analyze your clinical assay. Both platforms are available in 1- ,2-, and 4-channel configurations.
Thermo Scientific Transcend Duo UHPLC systems provide two independent channels with Vanquish pump technology to improve your productivity by saving time, reducing cost per sample, increasing capacity without added bench space, and using resources more efficiently.
Technical notes
Webinar
eBook
Posters
Case Studies
The Thermo Scientific VeriSpray PaperSpray Ion Source employs paper spray technology, a direct ionization technique that improves throughput, reduces cost per test and minimizes sample preparation. The technology is well suited for TDM, toxicology, and clinical research applications where time and cost per sample is a concern. The VeriSpray PaperSpray ion source uses simplified workflows that eliminate the need for more complex analytical techniques and enable the collection of quantitative and qualitative data by mass spectrometry.
Technical notes
Additional Ion Source resources
High-field asymmetric waveform ion mobility spectrometry Thermo Scientific (FAIMS) Pro Duo interface uses ion characteristics such as: charge state, shape, conformation, and size of the gas phase ion to reduce chemical noise and matrix interferences. These attributes provide assays with improved robustness and increased sensitivity.
Technical notes
Specification sheet
For General Lab Use Only – Not For Diagnostic Procedures