RNAlater™ Tissue Collection: RNA Stabilization Solution is the worldwide standard for the preservation of RNA in intact animal tissues, organs, and cells. RNAlater® eliminates the need to immediately process tissue samples or to freeze samples in liquid nitrogen for later processing. Here we demonstrate that RNAlater provides the same outstanding benefits when used with bacteria. It immediately stabilizes RNA expression profiles in both gram-positive and gram-negative species.
Integrity Analysis of RNA Isolated From RNAlater™ Stabilized Bacteria
Log phase Bacillus subtilis cells were harvested by centrifugation, resuspended in RNA later (or TE as a negative control), and incubated for four hours at room temperature or one week at 4°C. RNA was isolated from cells by bead-beating with zirconium/silica beads in RNAwiz™ (Ambion). RNA integrity was evaluated using the RNA 6000 LabChip and the Agilent 2100 bioanalyzer (Figure 1, A-D). Evaluation of the electropherograms obtained from the RNA later preserved samples, Figure 1C and 1D, clearly demonstrate that the quality of RNA recovered from B. subtilis cells stored in RNA later is indistinguishable from the positive control sample (Figure 1A). The RNA from B. subtilis stored in TE buffer for four hours is severely degraded (Figure 1B).
Figure 1. Bacterial RNA Stability. (A) Total RNA isolated from B. subtilis and processed immediately. The ratio of 23S:16S rRNA is 1.8. No RNA degradation is apparent. (B) Total RNA isolated from B. subtilis incubated in TE for 4 hr at 25ºC. Extensive degradation of 16S and 23S rRNAs is apparent. (C) Total RNA isolated from B. subtilis incubated in RNAlater for 4 hr at 25ºC. The ratio of 23S:16S rRNA is still 1.8. No RNA degradation is apparent. (D) Total RNA isolated from B. subtilis incubated in RNAlater for 1 week at 4ºC. The ratio of 23S:16S is still 1.8. No RNA degradation is apparent.
RNAlater™ is Effective at Preserving Bacterial RNA for Gene Expression Profiling With Arrays
To determine if storage of bacteria in RNAlater affects gene expression patterns, we compared the expression profile obtained with RNA isolated from B. subtilis that had been stored overnight in RNAlater at 25°C (Figure 2A) with the profile from RNA prepared from freshly harvested B. subtilis (Figure 2B). A comparison of these replicate arrays demonstrates that RNAlater is extremely effective at preserving gene expression profiles in bacteria. Similar results were obtained when arrays were probed with RNA prepared from B. subtilis cells that were stored in RNAlater for one week at 4°C. RNAlater is thus an ideal solution for maintaining the expression profile of bacterial mRNA when immediate RNA isolation is not possible or practical.
Ambion recommends the following protocol for preserving bacterial RNA using RNAlater. Cells should be collected by centrifugation and resuspended in RNAlater immediately after harvesting. Resuspend cell pellets from 2 ml of logarithmic phase cell culture in 0.5 ml - 1 ml of RNAlater. RNAlater-treated cells can be stored up to 8 hrs at 25°C or up to a week at 4°C. For longer storage, cells should be pelleted after incubation in RNAlater for one hour, flash frozen, and stored at -80°C.