">
Choose the orbital shaker that best fits your needs
Our portfolio of orbital shakers offers a wide selection of formats, features and capabilities that are ideal for nearly any application and lab environment.  Launch the interactive selection guide to choose the functionality and format that’s right for your work and your lab.
Comparison models
Open Air Shakers
CO2 Resistant Shakers
Benchtop Temperature Control Shakers
IntelliStack Incubator and
CO2 Incubator Shakers
Stackable Temperature Control Shakers
Temperature Ranges
Ambient
MaxQ 416 HP
Ambient
CO2
Resistant Shaker
Ambient
4°C to 60°C
4°C to 60°C
MaxQ 8000
5°C to 60°C
MaxQ 6000
Ambient -15°C to 80°C
Speed Ranges
15-525 rpm
MaxQ 416 HP
25-525 rpm
CO2 
Resistant Shaker
30-300 rpm
30-1200 rpm
15-525 rpm
30-350 rpm (25 mm)
30-300 rpm (50 mm)
MaxQ 8000
25-4000 rpm
MaxQ 6000
15-500 rpm
Orbit Sizes
0.4 in. (1.2cm)
.75 in. (1.9cm)
1 in. (2.5cm)
MaxQ 416 HP
1 in. (2.54 cm)
CO2 
Resistant Shaker
.75 in. (1.9 cm)
.12 in. (3 mm)
1 in. (2.5cm)
.1 in. (2.5cm)
2 in. (5.5 cm)
MaxQ 8000
1 in (2.5cm)
MaxQ 6000
.75in (1.9cm)
Weight Capacity
13 lb. (6kg) - 71 lb. (32kg)
MaxQ 416 HP
71 lb.
CO2 
Resistant Shaker
13 lb.
70 lb. (32kg)
44 lb. (20kg)
MaxQ 8000
71 lb. (32kg)
MaxQ 6000
35 lb. (15.9kg)
Platform Size Range
11 x 13 in. – 18 x 18 in.
MaxQ 416 HP
18 x 18 in.
18 x 24 in.
CO2 
Resistant Shaker
11.8 x 14.2 in.
18 x 18 in.
11 x 14 in. – 18 x 18 in.
35 x 22 in.
MaxQ 8000
18 x 30 in
MaxQ 6000
18 x 18 in
Compatible Accessories
Platforms
Clamps
Racks & Misc
Platforms
Clamps
Test Tube Racks
Plugs & Accessories
Platforms
Clamps
Racks & Misc
Clamps
Sticky Mats
Racks & Misc
Platforms
Clamps
Racks & Misc
">