Detect antigens and antibodies with traditional latex
Find your identification and confirmation solution from one of the largest portfolios of latex agglutination products in the world. With a wide range of organisms and organism types, you can find solutions that test from human samples including blood culture and colonies isolated on plated culture media.
Rapidly identify and differentiate strains of staphylococci by detecting the clumping factor and/or protein A characteristic of meticillin-resistant Staphylococcus aureus (MRSA). Use reliable latex testing to identify surface antigens associated with MSSA strains.
Isolate, identify and confirm streptococci with rapid, reliable results. Utilize acid or enzyme extraction for fast, qualitative detection of Lancefield groups A, B, C, D, F and G.
Detect, presumptively identify and/or confirm a variety of microbes from culture and serum, including E. coli, C. difficile, Helicobacter pylori, Listeria, Cryptococcus and Salmonella.
Rapidly detect and identify antigens to organisms, such as Haemophilus influenzae and Neisseria, from human body fluids, blood cultures or culture media.
Rapidly confirm and/or presumptively identify Escherichia coli serogroup O157:H7 and Shiga toxin-producing E. coli (STEC) with colonies isolated from plated media.
Screen for qualitative detection and semi-quantitative assessment of antibodies associated with a wide range of relevant organisms and illnesses including Syphilis, Lyme Disease, Toxoplasma gondii, rubella, and HSV in serum or plasma.
Detect Bacillis cereus or Clostridium perfringens enterotoxins and E. coli verocytotoxin VT1 and VT2 from fecal specimens, or detect staphylococcal toxic-shock syndrome toxin (TST) with a complete kit for Reversed Passive Latex Agglutination (RPLA).