">
Manual, semi-automated, or fully automated solutions across the AST workflow
The Sensititre System for MIC plate testing supports a mix of manual, semi-, and fully-automated solutions across the AST workflow, as illustrated in the Sensititre introductory video. You can choose which techniques and instruments best fit your lab’s testing routine.
To select Oxoid AST discs for manual testing, visit the Tests and Accessories page.
Which AST instruments are right for your workflow?
All Sensititre instruments and devices share common Sensititre System features:
  • Accurate, reliable, consistent, true MIC results
  • Applicable across clinical, veterinary, and surveillance domains
  • Seamless fit into an integrated workflow
  • Access to the latest antimicrobials
  • Compatibility with Sensititre custom AST plates
  • Premiere Thermo Scientific support and service
Use our Selection Guide to find the right instruments and devices for your AST workflow.
Selection guide
Select Plate
Sensititre Standard AST Plates
Choose from our wide selection of standard plates to suit your application, including Gram positive, Gram negative, fastidious, mycobacteria, yeast, and single-drug formats.
Learn more
Sensititre Custom AST Plates
Design your own plate from our selection of over 300 antimicrobials. We offer one of the widest, most up-to-date selections of antimicrobials, available in wide dilution ranges.
Learn more
Inoculum
Sensititre Nephelometer
A simple solution to standardize inoculum preparation and measure inoculum density.
Learn more
Inoculate
Sensititre 8-Channel Programmable Pipette
Quick and accurate manual inoculation of microtiter plates, with enhanced ergonomics.
Learn more
Sensititre AIM Automated Inoculation Delivery System
Automatically doses Sensititre plates and eliminates skipped wells and costly repeat tests.
Learn more
Incubate
Sensititre ARIS HiQ System
Individually incubates up to 100 Sensititre MIC, breakpoint, or identification plates, ensuring optimal growth conditions and eliminating repeat tests. Then automatically reads plates and prepares reports, improving efficiency without sacrificing accuracy.
Learn more
Read
Sensititre OptiRead Automated Fluorometric Plate Reading System
Uses fluorescence detection to automate Sensititre plate reading, delivering fast, accurate results. Directly links to Sensititre SWIN Software System to automate interpretation and reporting of results.
Learn more
Sensititre Vizion Digital MIC Viewing System
Captures and stores easy-to-read digital plate images for optimal manual reading and traceability. Connects to the Sensititre SWIN Software system for automated interpretation and reporting of results.
Learn more
Sensititre Manual Viewbox
Perform simple visual reads of your 96-well microtiter plates with our mirrored viewbox.
Learn more
Interpret
Sensititre SWIN Software System, Complete
Consolidates Sensititre results from manual and automated reading options on a single platform.
Learn more
Sensititre SWIN Epidemiology Module
Provides comprehensive reporting of AST results to help monitor antibiotic resistance patterns and support antimicrobial stewardship programs.
Learn more
Regulatory status varies by product and region. Please consult individual product pages.
">