Fixed and stained HDFn cells.
HDFn cells were fixed, permeabilized, and blocked using the Image-iT® Fixation/Permeabilization Kit (R37602). Golgi were labeled with an anti-golgin 97 antibody (A21270) followed by detection with an Alexa Fluor® 488 secondary (A11029). Cells were also stained with Alexa Fluor® 594 phalloidin (A12381) to label actin and NucBlue™ Fixed (R37606) to label nuclei. Finally, cells were mounted in ProLong® Gold antifade reagent.
Related Products
Related Images
A prometaphase muntjac skin fibroblast stained with Alexa Fluor® 350 phalloidin, an anti–a-tubulin antibody and an anti–cdc6 peptide antibody. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
Endothelial cells. Go ›
Mouse Anti-Alpha Tubulin Monoclonal Antibody (Cat. No. A11126) Go ›
1% Agarose gel containing 16S and 23S ribosomal RNA (rRNA). SYBR® Green II RNA gel stain. Go ›
Immunofluorescent staining using mouse anti-Golgin-97 monoclonal antibody Go ›
Phagocytosis in MMM Cells using pHrodo™ Red E. coli BioParticles® Conjugate® (Catalog # P35361) Go ›
Muntjac skin fibroblast labeled with rhodamine phalloidin. Go ›
CD335 (NKp46) Antibody (63335182) in RE Go ›
CD223 (LAG-3) Antibody (56223942) in TM Go ›
Increase in accuracy and precision using the Qubit® Go ›
Performance of the Qubit® dsDNA HS Assay Go ›
Muntjac fibroblast labeled with probes for actin and the nucleus. Go ›
Multicolor fluorescence analysis of muntjac fibroblasts. Go ›