Reprogramming somatic cells to induced pluripotent stem cells is a critical and potentially time intensive step in your stem cell research. That’s why Life Technologies offers choices in integration-free reprogramming technologies and services that fit your research and resource needs – you can do it yourself or let us do it for you.
Now available:Essential 6™ Medium for feeder-free reprogramming
Which reprogramming product or service is right for you?
Episomal iPSC Reprogramming Vectors* | Epi5™ Episomal iPSC Reprogramming Kit** | CytoTune®-iPS Sendai Reprogramming Kit | CytoTune®-iPS 2.0 Sendai Reprogramming Kit | |
---|---|---|---|---|
Order now | Order now | Order now | Order now | |
Description | Proven technology for viral-free iPSC generation | High efficiency viral-free system requiring no small molecules | Efficient generation of integration-free iPSCs from multiple somatic cell types | Highest efficiency integration-free reprogramming system |
Reprogramming Efficiency | 0.002–0.08% | 0.04–0.3% | 0.01-0.5% | 0.02 – 1.2% |
Genes Utilized | Thomson/Yamanaka factors | Yamanaka factors +Lin28 | Yamanaka factors | Yamanaka factors |
Method & Medium for Reprogramming Fibroblasts | Electroporation using Neon® in N2/B-27 Medium + small molecules | Electroporation using Neon® in N2/B-27 Medium | Add CytoTune® to cells in fibroblast medium | Add CytoTune® 2.0 to cells in fibroblast medium |
iPSC Growth Medium | Day 15: Transfer to Essential 8™ Medium | Day 15: Transfer to Essential 8™ Medium | Day 7: Transfer to Essential 8™ Medium or KnockOut™ Serum Replacement supplemented medium | Day 7: Transfer to Essential 8™ Medium or KnockOut™ Serum Replacement supplemented medium |
Genomic Integration-free | ✓ | ✓ | ✓ | ✓ |
Virus-free Reprogramming | ✓ | ✓ | ||
Blood Cell Reprogramming | ✓ | ✓ | ✓ | ✓ |
Transfection Products | Neon® Transfection System or similar device | Neon® Transfection System or Lipofectamine® 3000 | None | None |
Available in CellModel™ Services | ✓ | ✓ | ✓ | |
Viral Clearance | N/A | N/A | Up to 15 passages | Up to 5 passages |
Reporter Available | N/A | N/A | CytoTune®-EmGFP Sendai Fluorescence Reporter | CytoTune®-EmGFP Sendai Fluorescence Reporter |
**Designed by Dr. Okita in the laboratory of Professor Yamanaka at the Center for iPS Cell Research and Application (CiRA), Kyoto University