Your RNA sample may be of poor qualityYou may not have enough template sequenceYour reverse transcription may not be optimalYour assay design may not detect all of the splice variants for the gene of interest
For Research Use Only. Not for use in diagnostic procedures.