QuantiGene Singleplex Assay modules
The QuantiGene Singleplex Assay Kit is composed of several modules, each sold separately, depending on sample type of interest. Each of the modules is sold in a multiple sizes. The table below is a guideline for the required modules based on the sample type of interest.
QuantiGene Singleplex Assay Kit | Sample processing kit | Sample assessment kit | Housekeeping panel | QuantiGene Singleplex probe sets | |
---|---|---|---|---|---|
Cultured cells | Required | Cultured cells | n/a | Recommended | Required |
Whole blood or PAXgene blood | Required | Blood samples | n/a | Highly recommended | Required |
Fresh/frozen animal tissues | Required | Animal tissues | Optional | Highly recommended | Required |
FFPE tissue sections or blocks | Required | FFPE samples | Required | Highly recommended | Required |
Purified RNA | Required | n/a | n/a | Recommended | Required |
Video: How it works
For Research Use Only. Not for use in diagnostic procedures.