Life Technologies has the most complete collection of transfection reagents with exceptional performance for the delivery of siRNA, Stealth™ RNAi, RNAi vectors, DNA oligonucleotides, and RNA, in traditional or difficult-to-transfect cell lines.
The Mechanism of Cationic Lipid-Mediated Transfection
The positive surface charge of the liposomes also mediates the interaction of the nucleic acid and the cell membrane, allowing for fusion of the liposome/nucleic acid (“transfection complex”) with the negatively charged cell membrane. The transfection complex is thought to enter the cell through endocytosis. Endocytosis is the process where a localized region of the cellular membrane uptakes the DNA: liposome complex by forming a membrane bound/intracellular vesicle. Once inside the cell, the complex must escape the endosomal pathway, diffuse through the cytoplasm, and enter the nucleus for gene expression. Cationic lipids are thought to facilitate transfection during the early steps of the process by mediating DNA condensation and DNA/cellular interactions.
The principle of delivery using cationic lipid reagents thus differs from prior attempts to use neutral liposomes for transfections. With cationic lipid reagents, the DNA solution is not deliberately encapsulated within the liposomes, rather, the negatively charged DNA binds spontaneously to the positively charged liposomes, forming DNA-cationic lipid reagent complexes (Figure 1)
Some of the problems associated with traditional transfection methods like calcium phosphate coprecipitation, DEAE-dextran, polybrene and electroporation, include low efficiency of DNA delivery, poor reproducibility, cell toxicity, and inconvenience. However, cationic lipid reagent-mediated transfection yields high and previously unattainable transfection efficiencies in a wide variety of eukaryotic cells. It is simple to perform, and ensures consistently reproducible results. Moreover, a number of cell lines normally resistant to transfection by other methods transfect successfully using cationic lipid reagents.
Life Technologies Offers A Wide Range of Transfection Reagents for Lipid Transfection
- Click here for Plasmid DNA delivery
- Click here for siRNA delivery
References
- Chesnoy, S. and Huang, L. (2000) Annual Review of Biophysical Biomolecular Structure 29: 27-47.
- Hirko, A. et al. (2003) Current Medicinal Chemistry 10: 1185-1193.
- Liu, D. et al. (2003) Current Medicinal Chemistry 10: 1307-1315.