showcase.par.46476.image.0.0.1
Gibco® high-quality selection agents provide unique solutions for your research needs, such as dual selection and rapid stable cell line establishment.
Which Selection Antibiotic is Right for You?
Eukaryotic Selection Antibiotics
Selection Antibiotic Most Common Selection Usage Common Working Concentration Available Powder Sizes Available Liquid Sizes Order
BlasticidinEukaryotic and Bacteria1-20 µg⁄ml50 mg10x1 ml, 20 mlOrder Now
Geneticin (G-418)Eukaryotic100 - 200 µg ⁄ ml bacteria
200 – 500 µg ⁄ ml mammalian cells
1 g, 5 g, 10 g, 25 g20ml, 100mlOrder Now
Hygromycin BDual-selection Experiments and Eukaryotic200-500 µg ⁄ ml-20mlOrder Now
Mycophenolic AcidMammalian and Bacteria25 µg ⁄ ml500 mg-Order Now
PuromycinEukaryotic and Bacteria0.2 – 5 µg ⁄ ml-10 x 1 ml, 20mlOrder Now
ZeocinMammalian, insect, yeast, bacteria and plants50 – 400 µg ⁄ ml-8 x 1.25 ml, 50 mlOrder Now
Bacterial Selection Antibiotics
Selection Antibiotic Most Common Selection Usage Common Working Concentration Available Powder Sizes Available Liquid Sizes Order
Actinomycin DBacteria1 µg ⁄ ml5 mg, 10 mg-Order Now
Ampicillin Sodium SaltBacteria10 – 25 µg ⁄ ml200 mg-Order Now
BlasticidinEukaryotic and Bacteria50-100 µg ⁄ ml50 mg10x1ml, 20mlOrder Now
Carbenicillin, Disodium SaltAgrobacterium and E. coli100 – 500 µg ⁄ ml5 g-Order Now
Kanamycin SulfateBacteria100 µg ⁄ ml5 g, 25 g100mlOrder Now
Mycophenolic AcidMammalian and Bacteria25 µg ⁄ ml500 mg-Order Now
Neomycin SulfateBacteria50 µg ⁄ ml100 g-Order Now
Polymyxin B SulfateBacteria100 units / ml25 MU-Order Now
PuromycinEukaryotic and Bacteria0.2 – 5 µg ⁄ ml-10 x 1 ml, 20mlOrder Now
Streptomycin SulfateBacteria50 – 100 µg ⁄ ml-100 gOrder Now
ZeocinMammalian, insect, yeast, bacteria and plants75 – 400 µg ⁄ ml-8 x 1.25 ml, 50 mlOrder Now