Cell movement and location studies require specialized probes that are nontoxic to living cells and are available in a range of fluorescent colors to match instrument lasers and filters and to accommodate co-staining with antibodies or other cell analysis probes. Choose passively loaded dyes for short- and long-term tracking or fluorescent fusion proteins for cell tracking over 5 or more days.
Select products for cell tracking
On this page:
- Short-Term Cell Tracking
- Long-Term Cell Tracking
- Cell Tracking With Fluorescent Proteins
- Cell Tracking Selection Guides
The simplest processes for monitoring cell movement and location use tracking probes that pass through the membrane into the cell and become membrane-impermeant after loading. Ideal tracking dyes are passed to daughter cells for multiple generations but do not transfer to neighboring cells on contact. CellTracker™ dyes exhibit ideal properties—they are stable, nontoxic at working concentrations, well retained in cells, and brightly fluorescent at physiological pH. CellTracker™ dyes have a wide range of emission spectra for easy combination with antibody staining. | Rat basophilic leukemia (RBL) cells labeled in suspension with CellTracker™ Blue CMAC, CellTracker™ Green CMFDA or CellTracker™ Orange CMTMR prior to plating. |
Longer-term tracking of cell movement and location requires probes that are more resistant to photobleaching and are retained through more cycles of cell division. Qtracker® probes provide intense, stable signals for long-term (6–10 generations) tracking. The eight distinct probe colors are resistant to photobleaching and provide intense punctate fluorescence within the cell, with different emissions from a single excitation for easy combination with antibody staining. Qtracker® probes have a wide range of emission spectra for easy combination with antibody staining. | Multiplexing Qdot® probes, dyes, and fluorescent proteins. U2-OS cells were transduced with CellLight® Plasma Membrane-GFP (green), followed by labeling with Qtracker® 655 Cell Labeling Kit (magenta), and HCS NuclearMask™ Blue (blue). |
Transiently transduced cells express fluorescent fusion proteins at different locations within the cell for at least 5 days or up to 2 weeks in slow-growing populations. Labeled cells are easy to track, but not all cells within a population will express the fluorescent protein. CellLight® fluorescent proteins can be expressed in a variety of cell types including numerous transformed cell lines, primary cells, stem cells and neurons. | Porcine primary skeletal muscle cell transduced with CellLight® Nucleus-GFP fluorescent proteins. |
Short-term tracking requires rapid loading and typical retention for 3–6 generations or 72 hours.
Properties
Method of use | 15 minutes to 1 hour of cell loading |
Readout | Internalized probe gives bright fluorescence throughout the cell |
Range | Tracking extends for 3–6 generations or 72 hours |
Compatibility | Multiplex easily with antibody labeling after fixing |
Signal-to-noise | |
Photostability | |
Fluorescence microscopy | Yes |
HCS | Yes |
In vivo | |
Bibliography | Citations |
Selection Guide
CellTracker™ dye
|
Common filter set
|
Ex/Em (nm)
| Quantity | Cat. No. | |
---|---|---|---|---|---|
CellTracker™ Blue CMAC (7-amino-4-Chloromethylcoumarin) | CMAC | DAPI | 353/466 | 5 mg | C2110 |
CellTracker™ Blue CMF2HC (4-chloromethyl-6,8-difluoro-7-hydroxycoumarin) | Blue CMF2HC | DAPI | 371/464 | 5 mg | C12881 |
CellTracker™ Violet BMQC (2,3,6,7-tetrahydro-9- bromomethyl-1H,5H-quinolizino( 9,1-gh)coumarin) | Violet BMQC | DAPI | 415/516 | 5 x 0.1 mg | C10094 |
CellTracker™ Green CMFDA (5-chloromethylfluorescein diacetate) | Green CMFDA | FITC | 492/517 | 20 x 50 µg | C7025 |
CellTracker™ Orange CMRA | Orange CMRA | TRITC | 548/576 | 20 x 50 µg | C34551 |
CellTracker™ CM-DiI | CM-DiI | TRITC | 553/570 | 20 x 50 µg | C7000 |
CellTracker™ Red CMTPX | CMTPX | Rhodamine | 577/602 | 20 x 50 µg | C34552 |
CellTracker™ Deep Red | Deep Red | Cy®5 | 630/650 | 20 x 15 µg | C34565 |
Long-term tracking requires longer photostability and typical retention for 6–10 generations; loading time is less critical to experimental protocols.
Properties
Qtracker® 525 Cell Labeling Kit
| |
---|---|
Method of use | 1-hour cell loading using a targeting peptide |
Readout | Internalized probe gives intense punctate fluorescence within the cell |
Range | Tracking extends for 6–10 generations or 5–14 days |
Fixablility | Image cells live or after fixing |
Compatibility | Multiplex easily with antibody labeling |
Signal-to-noise | |
Photostability | |
Laser excitation | Excited optimally with UV up to 488 nm (also every wavelength below the emission wavelength) |
Fluorescence microscopy | Yes |
HCS | Yes |
In vivo | Yes |
Bibliography | Citations |
Selection Guide
Common filter set
|
Emission max (nm)
|
Quantity
| Cat. No. | |
---|---|---|---|---|
Qtracker® 525 Cell Labeling Kit | GFP or FITC | 525 | 1 kit | Q25041MP |
Qtracker® 565 Cell Labeling Kit | 565 | 1 kit | Q25031MP | |
Qtracker® 585 Cell Labeling Kit | TRITC-A | 585 | 1 kit | Q25011MP |
Qtracker® 605 Cell Labeling Kit | 605 | 1 kit | Q25001MP | |
Qtracker® 625 Cell Labeling Kit | Texas Red® | 625 | 1 kit | A10198 |
Qtracker® 655 Cell Labeling Kit | 655 | 1 kit | Q25021MP | |
Qtracker® 705 Cell Labeling Kit | Cy®5 | 705 | 1 kit | Q25061MP |
Qtracker® 800 Cell Labeling Kit | Cy®7 | 800 | 1 kit | Q25071MP |
Transduction with fluorescent proteins provides long-term tracking, but since not all cells are transformed some automated techniques are not supported.
Method of use | Transduction of cell line with fluorescent protein requires overnight incubation | ||
Readout | Up to 90% of cells express fluorescent protein in respective cell compartments | ||
Range | Expressed for ≥10 cell generations; 5 days to 2 weeks of tracking | ||
Dye | RFP | GFP | GFP |
Ex/Em (nm) | 555/584 | 488/510 | 488/510 |
Fixability | |||
Multiplexing | |||
Signal-to-noise | |||
Photostability | |||
Location | Nucleus | Cytoplasm | |
Fluorescence microscopy | Yes | ||
HCS | |||
In vivo | Yes | ||
Bibliography | Citations | ||
Quantity | 1 mL | 1 mL | 1 mL |
Cat. No. | C10603 | C10602 | B10383 |
Short-term tracking requires rapid loading and typical retention for 3–6 generations or 72 hours.
Properties
Method of use | 15 minutes to 1 hour of cell loading |
Readout | Internalized probe gives bright fluorescence throughout the cell |
Range | Tracking extends for 3–6 generations or 72 hours |
Compatibility | Multiplex easily with antibody labeling after fixing |
Signal-to-noise | |
Photostability | |
Fluorescence microscopy | Yes |
HCS | Yes |
In vivo | |
Bibliography | Citations |
Selection Guide
CellTracker™ dye
|
Common filter set
|
Ex/Em (nm)
| Quantity | Cat. No. | |
---|---|---|---|---|---|
CellTracker™ Blue CMAC (7-amino-4-Chloromethylcoumarin) | CMAC | DAPI | 353/466 | 5 mg | C2110 |
CellTracker™ Blue CMF2HC (4-chloromethyl-6,8-difluoro-7-hydroxycoumarin) | Blue CMF2HC | DAPI | 371/464 | 5 mg | C12881 |
CellTracker™ Violet BMQC (2,3,6,7-tetrahydro-9- bromomethyl-1H,5H-quinolizino( 9,1-gh)coumarin) | Violet BMQC | DAPI | 415/516 | 5 x 0.1 mg | C10094 |
CellTracker™ Green CMFDA (5-chloromethylfluorescein diacetate) | Green CMFDA | FITC | 492/517 | 20 x 50 µg | C7025 |
CellTracker™ Orange CMRA | Orange CMRA | TRITC | 548/576 | 20 x 50 µg | C34551 |
CellTracker™ CM-DiI | CM-DiI | TRITC | 553/570 | 20 x 50 µg | C7000 |
CellTracker™ Red CMTPX | CMTPX | Rhodamine | 577/602 | 20 x 50 µg | C34552 |
CellTracker™ Deep Red | Deep Red | Cy®5 | 630/650 | 20 x 15 µg | C34565 |
Long-term tracking requires longer photostability and typical retention for 6–10 generations; loading time is less critical to experimental protocols.
Properties
Qtracker® 525 Cell Labeling Kit
| |
---|---|
Method of use | 1-hour cell loading using a targeting peptide |
Readout | Internalized probe gives intense punctate fluorescence within the cell |
Range | Tracking extends for 6–10 generations or 5–14 days |
Fixablility | Image cells live or after fixing |
Compatibility | Multiplex easily with antibody labeling |
Signal-to-noise | |
Photostability | |
Laser excitation | Excited optimally with UV up to 488 nm (also every wavelength below the emission wavelength) |
Fluorescence microscopy | Yes |
HCS | Yes |
In vivo | Yes |
Bibliography | Citations |
Selection Guide
Common filter set
|
Emission max (nm)
|
Quantity
| Cat. No. | |
---|---|---|---|---|
Qtracker® 525 Cell Labeling Kit | GFP or FITC | 525 | 1 kit | Q25041MP |
Qtracker® 565 Cell Labeling Kit | 565 | 1 kit | Q25031MP | |
Qtracker® 585 Cell Labeling Kit | TRITC-A | 585 | 1 kit | Q25011MP |
Qtracker® 605 Cell Labeling Kit | 605 | 1 kit | Q25001MP | |
Qtracker® 625 Cell Labeling Kit | Texas Red® | 625 | 1 kit | A10198 |
Qtracker® 655 Cell Labeling Kit | 655 | 1 kit | Q25021MP | |
Qtracker® 705 Cell Labeling Kit | Cy®5 | 705 | 1 kit | Q25061MP |
Qtracker® 800 Cell Labeling Kit | Cy®7 | 800 | 1 kit | Q25071MP |
Transduction with fluorescent proteins provides long-term tracking, but since not all cells are transformed some automated techniques are not supported.
Method of use | Transduction of cell line with fluorescent protein requires overnight incubation | ||
Readout | Up to 90% of cells express fluorescent protein in respective cell compartments | ||
Range | Expressed for ≥10 cell generations; 5 days to 2 weeks of tracking | ||
Dye | RFP | GFP | GFP |
Ex/Em (nm) | 555/584 | 488/510 | 488/510 |
Fixability | |||
Multiplexing | |||
Signal-to-noise | |||
Photostability | |||
Location | Nucleus | Cytoplasm | |
Fluorescence microscopy | Yes | ||
HCS | |||
In vivo | Yes | ||
Bibliography | Citations | ||
Quantity | 1 mL | 1 mL | 1 mL |
Cat. No. | C10603 | C10602 | B10383 |
Resources | |
Molecular Probes® Handbook | BioProbes® articles |