Applied Biosystems TaqMan Gene Expression Assays represent the largest collection of predesigned assays in the industry with over 2.8 million assays across 32 eukaryotic species and numerous microbes. TaqMan Gene Expression assays enable you to get results fast with no time wasted optimizing SYBR Green primers and no extra time spent running and analyzing melt curves.
- Gold-standard specificity, sensitivity, and reproducibility
- Designed to detect virtually any transcript
- Best-coverage assays available to detect the highest number of transcript variants possible
- Performance guaranteed
Already know your target of interest?
Search for a pre-designed TaqMan Gene Expression Assay using the tool below.
Looking to design a custom TaqMan Gene Expression Assay?
Use our Custom Assay Design Tool.
Specifications
Species | TaqMan Gene Expression Assays are available for 32 species plus some common microbial species. |
Assay components | TaqMan Gene Expression Assays are delivered in a single tube containing the specific pre-formulated TaqMan Assay (TaqMan MGB probe labeled with FAM and forward and reverse primers). |
Assay suffix nomenclature | _s1: an assay with primers and probes designed within a single exon. Such assays, by definition, detect genomic DNA. _g1: an assay that may detect genomic DNA, with primers and probe that may be within a single exon _m1: an assay with a probe that spans an exon junction _mH, _sH, and _gH: an assay that is designed to a transcript belonging to a gene family that has high sequence homology |
Available sizes | TaqMan Gene Expression Assays are available in numerous dye and size combinations:
|
Shipping and storage | Shipped at ambient temperature. |
Data analysis tools
- Connect cloud-based platform—free to sign up
- Desktop software—available through the Support Center for your specific qPCR instrument
- ExpressionSuite Software—free to download