Integrated all-in-one vacuum concentrator solutions
By combining the three major components of a concentration system into a single housing and making the components ritually maintenance -free, the SPD Integrated SpeedVac kits take sample concentration to a higher level. Completely self contained, the SPD1030 and SPD2030 SpeedVacs provide a "plug and play" convenient setup with uncomplicated controls, with built in sample evaporation programs and display. All integrated kits are UL Listed (US & Canadian requirements), CE marked, China ROHS certified.
SpeedVac Selection Guide
The SPD Integrated SpeedVac systems combines 4 major components into a single housing:
  1. SpeedVac concentrator
  2. 4L -50°C refrigerated vapor trap
  3. Vacuum controller
  4. Oil-Free ETFE Diaphragm Pump
View all SpeedVac rotors
Choose your Integrated SpeedVac Kit
DNA130
SPD1030
SPD2030
Capacity36 x 1.5mL
2 shallow well plates
120 x 1.5mL
4 flasks
10 15mL tubes
2 shallow-well plates
(Other tube & vial formats)
200 x 1.5mL
8 flasks
52 15mL tubes
8 deep-well plates
(Other tube & vial formats)
Compatible rotor types52030
Components includedOil free pumpOil free pump, -50°C cold trapOil free pump, -50°C cold trap
Number of programs3 (customizable by application)3 preset, 3 user defined3 preset, 3 user defined
Preset by solvent typeWater, ethanolWater, low boiling solvents, HPLCWater, low boiling solvents, HPLC
Type of solventsEthanol, water, PCR buffersEthanol, water, ammonium hydroxide, acetonitrileEthanol, water, ammonium hydroxide, acetonitrile
Product dimensions (WxDxH)11.5 x 25 x 12 in. (29 x 64 x 31cm)25 x 26 x 16 in. (64 x 66 x 41cm)31 x 28 x 19 in. (79 x 71 x 49cm)
Type of lidAcrylicRadiant glassRadiant glass
Available kits
Kit with rotor
DNA130-115: 115V/50Hz
DNA130-230: 230V/50Hz
  • Integrated SpeedVac system
  • 36 × 1.5mL rotor (RD36)
Kit without rotor
SPD1030A-115: 115V/60Hz
SPD1030A-230: 230V/50Hz
  • Integrated SpeedVac system
  • CryoCool™ 1 L heat transfer fluid
  • Glass condensation Flask (GCF400)
Kit with rotor
SPD1030P1-115: 115V/50Hz
SPD1030P1-230: 230V/50Hz
  • Integrated SpeedVac system
  • 40 × 1.5mL rotor (RH40-11)
  • CryoCool™ 1 L heat transfer fluid
  • Glass condensation Flask (GCF400)
Kit without rotor
SPD2030A-220: 220V/60Hz
SPD2030A-230: 230V/50Hz
  • Integrated SpeedVac system
  • CryoCool™ 1 L heat transfer fluid
  • Glass condensation Flask (GCF400)
Kit with rotor
SPD2030P1-220: 220V/50Hz
SPD2030P1-230: 230V/50Hz
  • Integrated SpeedVac system
  • 200 × 1.5mL rotor (RH200-12)
  • CryoCool™ 1 L heat transfer fluid
  • Glass condensation Flask (GCF400)