Immunofluorescent analysis of CFTR using an antibody recognizing CFTR shows staining in WiDr colon carcinoma cells. CFTR staining (green), F-actin staining with Phalloidin (red), and nuclei with DAPI (blue) is shown. Cells were grown on chamber slides and fixed with formaldehyde prior to staining. Cells were probed without (negative control) or with or a CFTR monoclonal antibody (Cat. No. MA1-935) at a dilution of 1:100–1:200 overnight at 4OC, washed with PBS and incubated with a DyLight 488−conjugated secondary antibody (Cat. No. 35552 for GAR; Cat. No. 35503 for GAM). Images were taken at 60x magnification.
Endosomes are membrane-bound vesicles formed through the invagination of the plasma membrane. They are part of the endocytic membrane transport pathway from the plasma membrane to the lysosome and allow molecules to be sorted before reaching the lysosome. Molecules internalized at the plasma membrane can be transported to lysosomes for degradation, or recycled to the plasma membrane. Endosomes can also transport molecules originating from the Golgi on to lysosomes or recycle them back to the Golgi.
Endosomes are found in three forms: early endosomes, late endosomes, and recycling endosomes. The early endosomes develop into late endosomes, which then fuse with lysosomes. Each endosome form has distinct morphology and specific markers.
Endosomal marker antibodies can aid in the study of the morphology and dynamics of the endosome. Endosomal marker antibodies can also help elucidate the role or roles a protein may play in a number of tasks that are centered in or influenced by the endosome. In addition, endosomal marker antibodies can track cell surface receptors that induce receptor-mediated endocytosis and are often studied as transporters of targeted delivery of enzymes, drugs, toxins, or DNA for therapeutic treatments. Invitrogen endosomal marker antibodies are designed to dependably detect the key endosomal targets. Each antibody is validated for use in various applications. Key endosomal marker targets include:
Applications
Immunohistochemistry performed on normal biopsies of deparaffinized human pancreas tissue. To expose target proteins, heat-induced antigen retrieval was performed using 10 mM sodium citrate (pH 6.0) buffer, microwaved for 8–15 minutes. Following antigen retrieval tissues were blocked in 3% BSA-PBS for 30 minutes at room temperature. Tissues were then probed at a dilution of 1:200 with a mouse monoclonal antibody recognizing CFTR (Cat. No. MA1-935) or without primary antibody (negative control) overnight at 4°C in a humidified chamber. Tissues were washed extensively with PBST and endogenous peroxidase activity was quenched with a peroxidase suppressor. Detection was performed using a biotin-conjugated secondary antibody and SA-HRP, followed by colorimetric detection using DAB. Tissues were counterstained with hematoxylin and prepped for mounting.
Resources
Downloads
Product selection guides
Learning and support