Environmental monitoring is essential for contamination control in pharmaceutical manufacturing, enabling early detection of risks through continuous monitoring of air quality, surface cleanliness, and water systems. This proactive approach allows swift corrective actions to prevent product contamination and ensure quality standards. Regular monitoring assesses contamination levels, informs adjustments to control measures, ensures regulatory compliance, and enhances patient safety.
Sampling methods in environmental monitoring within pharmaceutical manufacturing include using standardized procedures and validated techniques to ensure reliable and accurate results. Air sampling should be performed with calibrated active air samplers or settle plates placed strategically throughout the cleanroom. Passive air sampling can be useful in critical areas where active sampling could be intrusive and a hazard to the aseptic operation.
Surface and personnel sampling should be done periodically, especially after critical interventions, while minimizing process.
Selection of monitoring locations should be guided by risk assessments and ensure representation of different processes and environmental conditions.
Identified locations include high-risk areas that are susceptible to contamination and critical control points (CCPs) where monitoring is essential for preventing and detecting contamination.
Monitoring locations should be regularly reviewed and adjusted based on trend analysis and changes in production processes to maintain effective contamination control and compliance with quality standards.
Establishing alert and action limits is essential for interpreting environmental monitoring data and triggering appropriate responses to deviations from predefined standards. Alert limits indicate a potential deviation from normal operating conditions, prompting further investigation and corrective actions. Action limits represent thresholds beyond which corrective actions must be taken to prevent product contamination or quality issues. Setting scientifically justified alert and action limits based on risk assessments and historical data ensures that environmental monitoring programs are effective in identifying and addressing contamination risks in a timely manner.
Aseptic process simulations, or media fills, are a critical component of the overall environmental monitoring program. They involve using a growth medium in place of the actual product to simulate the aseptic filling process, ensuring that the manufacturing environment, procedures, and personnel can maintain sterility. These simulations help detect potential contamination risks, validate aseptic techniques, and demonstrate compliance with regulatory requirements.
Quality culture media is at the core of effective media fills. Explore our range which includes animal origin and animal origin free media both in dehydrated media and bioprocess bag formats.
Before using media for product testing, it should undergo quality control testing in accordance with relevant Pharmacopeial standards. Additionally, media intended for environmental monitoring and APS should be tested for growth promotion prior to use, employing a scientifically justified selection of reference microorganisms, including representative local isolates.
Complete your workflows with our ISO 17034 accredited and fully characterized ATCC® Licensed Derivative strains quality control microorganisms for growth promotion testing.
*The ATCC Licensed Derivative® Emblem, the ATCC Licensed Derivative® word mark and the ATCC catalog marks are trademarks of ATCC. Remel Inc. is licensed to use these trademarks and to sell products derived from ATCC® cultures. The identity, purity, and authenticity of the Licensed Products are exclusively the responsibility of Remel Inc., and not ATCC.
Thermo Fisher Scientific Microbiology is now an official accredited reference material (RM) producer meeting the requirements of ISO 17034, providing trusted reference material for over 400 of our Thermo Scientific Culti-Loops Qualitative Quality Control Organisms and 25 Thermo Scientific Quanti-Cult Plus Quantitative Quality Control Organisms.
For Laboratory Use Only. Not for use in diagnostic procedures.