Monitor microbial air contamination with our range of air samplers for use in the pharmaceutical and food production areas, clinical settings, or for environmental monitoring.
Monitor microbiological air quality in controlled pharmaceutical and food processing environments with the Thermo Scientific™ Oxoid™ Air Sampler. This robust, high specification air sampler uses proven impaction technology for reliability in quality assurance and infection control applications.
Based on a cyclonic technology combined with a high air flow rate, Coriolis μ offers efficient particle collection in 10 minutes. Biological particles such as toxins, viruses, bacteria, moulds, pollens, and spores are collected and concentrated in a liquid, ready to be analysed with a range of downstream experiments and biological analysis methods such as culturing, immunological tests, next generation sequencing, spectrometry, and more.
Spin Air samplers allow a compact, portable, simple solution to air sampling. These units are compact and devised for easy transportation to enable sampling to take place anywhere needed.