Water Microbiology Testing
Microbiology products for the isolation, identification and enumeration of waterborne microorganisms to meet your testing needs.
Our range of products for water testing includes dehydrated and prepared media, membrane filters, qPCR assays for Legionella, quality control organisms, as well as the equipment you need to deliver reliable results.
  • Safe: media formulations that are compliant with ISO Standards for the testing of water
  • Conform to ISO standard: quality control testing methods for culture media are accredited in accordance with ISO 17025:2017
  • Proven and certified: all Certificates of Analysis (CoAs) for culture media confirm testing in accordance with the ISO 11133:2014 standard, including membrane filter testing
  • Support: our team of microbiology experts is at hand to help with your technical queries
Request a call
Featured products
qualyfast-range-278x140
QuantStudio5-278x140
GVPC-plate-Legionella-media-278x140
NEW Legionella anisa QC testing strain available
NEW Legionella anisa QC testing strain available
To support your laboratory’s water testing processes, we have added Thermo Scientific™ Culti-Loops™ Quality Control Legionella anisa NCTC 11974 (WDCM 00106) to our quality control testing organism range. Culti-Loops Quality Control organisms enable quick and safe preparation of cultures for QC testing with ready-to-use, disposable inoculation loops containing gel-stabilised micro-organisms.
Legionella anisa is the second most often isolated Legionella species from water samples, after Legionella pneumophila.
Find out more
Detection of microorganisms: ISO 6222:1999
Water sample
Poured plate method with Yeast Extract Agar
68±4 h at 20±2 °C and 44±4 h at 36±2 °C
Count all colonies formed
MethodMediaProductFormatProduct Code
Pour plateYeast Extract AgarPlate Count Agar for water testing (ISO)500 g
Dehydrated culture media
CM1012B
Plate Count Agar10 x 100 mL bottles
Prepared media
BO0055M
10 x 200 mL bottles
Prepared media
BO0055R
Detection of Escherichia coli / Coliform bacteria: ISO 9308-1:2014
Water sample
Membrane filtration, using filter on surface of CCA Agar
36±2 ºC 21±3 h
Count all rose colonies as coliforms
Count all dark blue to purple colonies as E.coli
Test oxidase with freshly prepared reagent
Negative: No blue colonies
Confirmed E.coli
> Count all Oxidase-negative bacteria as coliforms
> Count all Oxidase-positive bacteria as non-coliforms
MethodMediaProductFormatProduct code
Membrane filtrationCCA AgarThermo Scientific™ Chromogenic Coliform Agar500 g
Dehydrated culture media
CM1205B
10 x 90 mm plates
Prepared media
PO5318A
10 x 55 mm plates
Prepared media
PO5428J
Oxidase testTryptone Soya Agar (TSA)CASO AGEN (TSA)500 g / 5 kg
Dehydrated culture media
CM0131B/T
10 x 90 mm plates
Prepared media
PO5012A
Oxidase ReagentThermo Scientific™ BactiDrop™ Oxidase Reagent50 vials
Dehydrated culture media
R21540
10 x 90 mm plates
Prepared media
PO5321A
Thermo Scientific™ Microbact™ Oxidase Strips50 strips
Dehydrated culture media
MB0266A
Detection of Enterococci: ISO 7899-2:2000
Water sample
Membrane filtration, using filter on surface of Slanetz and Bartley Agar
36±2 °C 44±4 h
Count all convex red, brown or rose colonies
Put filter onto pre-warmed 44°C
Bile Aesculine Azide Agar
44±0.5°C 2 h
Enumerate all brown colonies with brown halos as intestinal Enterococci
MethodMediaProductFormatProduct code
Membrane filtrationSlanetz Bartley AgarSlanetz Bartley Agar500 g
Dehydrated culture media
CM0377B
10 x 90 mm plates
Prepared media
PO5018A
10 x 55 mm plates
Prepared media
PO5410J*
10 x 55 mm plates
Prepared media
PO5423J*
ConfirmationBile Aesculine Azide Agar
Enterococcus Selective Medium
(Bile Aesculine Azide Agar)1
10 x 90 mm plates
Prepared media
PO5062A
or Kanamycin Aesculin Azide Agar Base2500g
Dehydrated culture media
CM0591B
10 x 90 mm plates
Prepared media
PO5059A
* Minimum order quantity required.
1. This medium is slightly different to the medium formulation according to ISO 7899-2:2000. The medium has additional sodium citrate and 20 g/l ox bile instead of 10 g/l and 0.55 g/l sodium azide instead of 0.15 g/l.
2. This medium is slightly different to the medium formulation according to ISO 7899-2:2000. It contains sodium citrate instead of ox bile and Kanamycin.
Detection of Pseudomonas aeruginosa: ISO 16266:2006
Water sample
Membrane filtration, using filter on surface of Pseudomonas CN Agar
36±2 °C 44±4 h*
Inspection under UV light
(360±20 nm)
Inspection under natural light
Inspection under natural light
Fluorescence colonies: presumptive Pseudomonas aeruginosa
Blue green (Pyocyanine forming) colonies: confirmed Pseudomonas aeruginosa
Red brown colonies: presumptive Pseudomonas aeruginosa
Subculture to Nutrient Agar
Subculture to Nutrient Agar
36±2°C 22±2 h
36±2°C 22±2 h
Add Nessler´s Reagent ammonia development (red-yellow precipitation)
Oxidase test: oxidase positive
Confirmed Pseudomonas aeruginosa
Acetamide Solution
King´s B Medium
36±2°C 22±2 h
36±2°C up to 5 days
Add Nessler´s Reagent ammonia development (red-yellow precipitation)
Fluorescence under UV light
Confirmed Pseudomonas aeruginosa
* Minimum order quantity required.
MethodMediaProductFormatProduct code
Membrane filtrationPseudomonas Selective Agar / CN AgarPseudomonas Centrimide Selective Agar10 x 90 mm plates
Prepared media
PO5076A
10 x 55 mm plates
Prepared media
PO5413J*
Pseudomonas Agar Base500 g
Dehydrated culture media
CM0559B
10 tubes
Dehydrated culture media
SR0102E
Confirmation
FluorescenceKing´s B Media   
Hydrolysis of acetamide to ammoniaAcetamide Solution Nessler Reagent   
Oxidase TestNutrient AgarNutrient Agar500 g
Dehydrated culture media
CM0003B
Oxidase ReagentBactiDrop Oxidase Reagent50 vials
Dehydrated culture media
R21540
Microbact Oxidase Strips50 strips
Dehydrated culture media
MB0266A
* Minimum order quantity required.
Detection of Clostridium perfringens (including spores): ISO 14189:2013
Membrane filtration of 100 mL onto TSC-Agar* (if necessary, dilute sample)
44±1°C, anaerobic 21±3 h
Enumerate the presumptive colonies which show black or grey to yellow brown staining within 30 minutes of opening
Count all black colonies as Clostridium perfringens
1-10 black or grey to yellow-brownish colonies
>10 black or grey to yellow-brownish colonies
Subculture all presumptive colonies onto blood agar plates 
Subculture 10 different presumptive colonies onto blood agar plates
36±2°C, anaerobic 21±3 h
36±2°C, anaerobic 21±3 h
Put colonies that grew under anaerobic conditions onto filter paper and add 2-3 drops of phosphateacid. All colonies that turn purple within 3-4 minutes are confirmed positive.
*ATTENTION: Alternatively, a thin layer (approximately 5 mL to 10 mL) of molten TSC Agar (TV5204G TSC Agar Base), as an overlay on the filter can be used. Allow to solidify before anaerobic incubation. This method may enhance the blackening of the colonies. Medium without cycloserine; e.g. TV5204G TSC Agar Base, 20 mL.
Melt TSC Agar in a water bath and put melted agar onto filter on the plate:
Melt agar in a water bath at 95°C for approx. 10-15 minutes
Cool medium to 45°C
Shake tube carefully and pour a thin layer of molten agar
MethodMediaProductFormatProduct code
Membrane filtrationTSC AgarTSC Selective Agar10 x 90 mm plates
Prepared media
PO5315A
TSC Agar Base +500 g
Dehydrated culture media
CM0587B
50 tubes
Prepared media
TV5204G
TSC Selective Supplement10 tubes
Dehydrated culture media
SR0088E
10 x 100 mL bottles
Prepared media
BO0634M
10 tubes
Prepared media
SR0088E
Anaerobic atmosphere Thermo Scientific™ AnaroGen™ System10 sachets
AN0025A
Thermo Scientific™ AnaroJar™ 2,5 L Anaerobic System1 unit
AG0025A
Thermo Scientific™ AnaeroBox™ Rectangular Anaerobic System1 unit
AB0025A
Detection of Legionella: ISO 11731:2017
Sample preparation:
Heat treated: Put sample into water bath for 30+/-2 minutes at 50 °C
Acid treated: 1 part sample to 9 parts of acid buffer. Mix well and leave it for 5+/- 0.5 minutes.
Acid treated filter: Put 30 mL of acid solution onto the membrane filter. Leave it for 5 +/- 0.5 minutes and remove the solution by filtration. Wash with 20 mL of either sterile Aqua dest., Pages Saline, Ringer Solution or PBS.
Samples with high concentration of Legionella species and low concentration of interfering organisms2
Samples with low concentration of Legionella species and low concentration of interfering organisms2
0.1-0.5 mL of the sample onto BCYE 
0.1-0.5 of the sample onto BCYE with AB
Membrane filtration of 100 mL sample1.
Place one untreated membrane filter onto BCYE Agar; Place membrane filter with acid solution on one or more selective media: BCYE Agar with AB or GVPC Agar or MWY Agar
0.1-0.5 mL of the sample; each untreated, heat treated and acid treated from the membrane filtration with washing procedure3 onto BCYE Agar and on one or more selective media: BCYE Agar with AB or GVPC Agar or MWY Agar
36 ± 2 °C, humid atmosphere, 7-10 days
Subculture at least 3 typical Legionella colonies from each plate and inoculate on BCYE Agar without Cystein and BCYE Agar
36 ± 2 °C, humid atmosphere, 2-5 days
Colonies that grow on BCYE but not on BCYE without cystein are considered to be Legionella3
  1. 10 – 1000 mL sample volume according to ISO 11731-2.
  2. The choice of the method for the enumeration of Legionella species depends on the origin/characteristics of the sample and the reason for sampling or investigation. A decision matrix can be found in the ISO 11731:2017, Annex J.
  3. Further information, please see ISO 11731:2017.
MethodMediaProductFormatProduct code
Sample preparationAcid BufferLegionella Acid Buffer Solution6 x 1 L bottles
Prepared media
GFB01
Direct and membrane filtrationLegionella GVPC AgarLegionella GVPC Selective Agar10 x 90 mm plates
Prepared media
PO5074A
10 x 90 mm plates
Prepared media
PO0245A
Legionella CYE Agar-Base +500 g
Dehydrated culture media
CM1203B
Legionella BCYE Supplement +10 tubes (500 mL)
Dehydrated culture media
SR0251C
Legionella GVPC Selective Supplement10 tubes (500 mL)
Dehydrated culture media
SR0252E
Legionella GVPC Agar with antibioticsLegionella BCYE Agar with antibiotics10 x 90 mm plates
Prepared media
PO5325A
Legionella BCYE AgarLegionella BCYE Agar10 x 90 mm plates
Prepared media
PO5072A
Legionella CYE Agar Base +500 g
Dehydrated culture media
CM1203B
Legionella BCYE Supplement10 tubes (500 mL)
Dehydrated culture media
SR0251C
Legionella MWY AgarLegionella MWY Agar10 x 90 mm plates
Prepared media
PO5071A
Legionella CYE Agar Base +500 g
Dehydrated culture media
CM1203B
Legionella BCYE Supplement +10 tubes (500 mL)
Dehydrated culture media
SR0251C
Legionella MWY Selective Supplement10 tubes (500 mL)
Dehydrated culture media
SR0255B
SubcultureLegionella BCYE Agar without CysteineLegionella BCYE Agar without Cysteine10 x 90 mm plates
Prepared media
PO5028A
10 x 90 mm plates
Prepared media
PO0255A*
Legionella CYE Agar Base +500 g
Dehydrated culture media
CM1203B
Legionella BCYE without Cysteine Supplement10 tubes (100 mL)
Dehydrated culture media
SR0253A
Legionella BCYE AgarLegionella BCYE Agar10 x 90 mm plates
Prepared media
PO5072A
Legionella CYE Agar Base +500 g
Dehydrated culture media
CM1203B
Legionella BCYE Supplement10 tubes (500 mL)
Dehydrated culture media
SR0251C
* Minimum order quantity required.
Detection of Legionella: qualyfast® qPCR
Detection of Legionella qualyfast qPCR workflow
ProductFormatOrder code
qualyfast Reagents and PCR Kits
qualyfast Free DNA Inactivator200 reactionsD401009
qualyfast DNA Extraction Kit I clear water24 reactionsD101001
48 reactionsD201001
96 reactionsD301001
qualyfast DNA Extraction Kit II dirty water48 reactionsD211001
96 reactionsD311001
qualyfast Legionella PCR Assay24 reactions
(3x 8-tube strips)
A101013
48 reactions
(6x 8-tube strips)
A201013
96 reactions
(12x 8-tube strips)
A301013
qualyfast Legionella Quantification Standards8 tests
(4 standards each in an 8-tube strip)
A501013
QuantStudio 5 Food Safety PCR System
Applied Biosystems™ QuantStudio 5 Food Safety Real-Time PCR System, 0.1 mL Block, LaptopSystemA36328
Applied Biosystems QuantStudio 5 Food Safety Real-Time PCR System, 0.1 mL Block, DesktopSystemA36320
Service & Support Packages
Applied Biosystems Assurance Service Plan
additional 1 year warranty cover with instrument loan
1 year coverSCQS5FAST
Filter Funnels and Membranes
Thermo Scientific™ Nalgene™ Membrane Filters are cellulose nitrate membranes that are certified for microbiological QC testing and analysis of water.
ProductSize of QuantityCatalog Number
Nalgene Disposable Analytical Funnels equipped with membrane filter
Sterile Analytical Filter Units, 100 mL, 47 mm, 0.45 μm, white/black50 piecesNG145-0045
Sterile Analytical Filter Units, 250 mL, 47 mm, 0.45 μm, white/black50 piecesNG145-2045
Sterile Analytical Filter Units, 100 mL, 47 mm, 0.45 μm, grey/black50 piecesNG147-0045
Sterile Test Filter Funnel, 250 mL, 0.45 uL50 piecesNG147-2045
Membrane filter
Membrane Filter for water testing, sterile, CN, 47 mm, 0,45 μm, white/black100 piecesNG0205-4045
Membrane Filter for water testing, sterile, CN, 47 mm, 0,45 μm, grey/black100 piecesNG0210-6045
Accessories
Vacuum Manifold1 pieceNG0345-0001
Filter Stopper, non-sterile, no. 8 rubber3 piecesNG0396-0080
Filter Forceps, bent tip1 pieceNG0399-0001
Filter Forceps, straight tip1 pieceNG0399-0002
Filter Funnel Adapter, non-sterile25 piecesNG0397-0010
Vacuum Gasket, non-sterile thermoplastic elastomer6 piecesNG0395-0708
Nalgene Reusable Analytical Filters and accessories
Filter Funnels with Clamp, 250 mL1 pieceNG0315-0047
Filter Holders with Receiver, 500 mL, 500 mL4 piecesNG0300-4000
Filter Holders with Receiver, 250 mL, 250 mL4 piecesNG0300-4050
Filter Holders with Receiver, 500 mL, 1000 mL4 piecesNG0300-4100
Filter Holders with Funnel, 250 mL1 pieceNG0310-4000
Filter Holders with Funnel, 500 mL1 pieceNG0310-4050
Reusable Bottle Top Filters, 250 mL1 pieceNG0320-2545
Reusable Bottle Top Filters, 500 mL1 pieceNG0320-5033
Reusable Bottle Top Filters, 500 mL1 pieceNG0320-5045
Quality Control organisms: ISO 11133:2014
ProductSample numbersCatalog Number
Escherichia coli QC Organisms for testing of CCA Agar
Escherichia coli ATCC® 8739™*WDCM 00012R4607085
Escherichia coli ATCC® 25922™*WDCM 00013R4607050
Enterobacter aerogenes ATCC® 13048™*WDCM 00175R4607080
Pseudomonas aeruginosa ATCC® 27853™*WDCM 00025R4607060
Pseudomonas aeruginosa ATCC® 9027™*WDCM 00026R4605210
Enterococcus faecalis ATCC® 19433™*WDCM 00009R4601990
Enterococcus faecalis ATCC® 29212™*WDCM 00087R4607030
Enterococci QC Organisms for testing of Slanetz and Bartley Agar
Escherichia coli ATCC® 8739™*WDCM 00012R4607085
Escherichia coli ATCC® 25922™*WDCM 00013R4607050
Staphylococcus aureus subsp. aureus ATCC® 6538™*WDCM 00032R4607016
Staphylococcus aureus subsp. aureus ATCC® 25923™*WDCM 00034R4607010
Enterococcus faecalis ATCC® 29212™*WDCM 00087R4607030
Enterococcus faecalis ATCC® 19433™*WDCM 00009R4601990
Pseudomonas aeruginosa QC Organisms for testing of Pseudomonas CN Agar
Pseudomonas aeruginosa ATCC® 27853™*WDCM 00025R4607060
Pseudomonas aeruginosa ATCC® 9027™*WDCM 00026R4607060
Pseudomonas aeruginosa ATCC® 10145™*WDCM 00024R4607065
Escherichia coli ATCC® 8739™*WDCM 00012R4607085
Escherichia coli ATCC® 25922™*WDCM 00013R4607050
Enterococcus faecalis ATCC® 19433™*WDCM 00009R4601990
Enterococcus faecalis ATCC® 29212™*WDCM 00087R4601990
Clostridium perfringens QC Organisms for testing of TSC Agar
Bacillus subtilis ATCC 6633WDCM 00003R4601221
QC organisms for testing GCVP Agar (Legionella method)
Legionella pneumophila ATCC® 33152™*WDCM 00107R4603950
Legionella anisa NCTC 11974™WDCM 00106R4601315
Enterococcus faecalis ATCC® 19433™*WDCM 00009R4601990
Enterococcus faecalis ATCC® 29212™*WDCM 00087R4607030
Pseudomonas aeruginosa ATCC® 27853™*WDCM 00025R4607060
Pseudomonas aeruginosa ATCC® 9027™*WDCM 00026R4605210
Escherichia coli ATCC® 8739™*WDCM 00012R4607085
Escherichia coli ATCC® 25922™*WDCM 00013R4607050
Prepared Media for the Quality Control testing according to ISO 11133:2014
CASO Agar (ISO 11133) PO5321A
* The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Remel Inc. is licensed to use these trademarks and sell products derived from ATCC® cultures. The identity, purity, and authenticity of the Licensed Products are exclusively the responsibility of Remel Inc., and not ATCC.